Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14039832-14040073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL27RA
Entrez Gene ID
9466
Gene Name
interleukin 27 receptor subunit alpha
Gene Aliases
CRL1, IL-27RA, IL27R, TCCR, WSX1, zcytor1
UniGene
Hs.132781
Gene Chromosome Location
Chr.19: 14031740-14052919 on Build GRCh38
Transcripts
NM_004843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000203 c.521C>A p.A174E Chr.19: 14039897-14039897 on Build GRCh38
3281534 c.521C>T p.A174V Chr.19: 14039897-14039897 on Build GRCh38
1711894 c.530C>T p.T177I Chr.19: 14039906-14039906 on Build GRCh38
4801932 c.477T>C p.H159H Chr.19: 14039853-14039853 on Build GRCh38
3891649 c.527G>A p.W176* Chr.19: 14039903-14039903 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1046851728 Chr.19: 14039968-14039968 on Build GRCh38
rs768973217 Chr.19: 14039856-14039856 on Build GRCh38
rs757216991 Chr.19: 14039917-14039917 on Build GRCh38
rs763916613 Chr.19: 14039899-14039899 on Build GRCh38
rs149150973 Chr.19: 14039882-14039882 on Build GRCh38
rs750374493 Chr.19: 14039931-14039931 on Build GRCh38
rs764852881 Chr.19: 14039890-14039890 on Build GRCh38
rs552269402 Chr.19: 14040006-14040006 on Build GRCh38
rs369206630 Chr.19: 14039951-14039951 on Build GRCh38
rs765418656 Chr.19: 14039968-14039970 on Build GRCh38
rs776197914 Chr.19: 14039881-14039881 on Build GRCh38
rs372276918 Chr.19: 14039945-14039945 on Build GRCh38
rs577104639 Chr.19: 14040024-14040024 on Build GRCh38
rs555116302 Chr.19: 14039937-14039937 on Build GRCh38
rs747540021 Chr.19: 14039956-14039956 on Build GRCh38
rs756141524 Chr.19: 14039933-14039933 on Build GRCh38
rs186420427 Chr.19: 14039942-14039942 on Build GRCh38
rs58817586 Chr.19: 14039947-14039947 on Build GRCh38
rs772509590 Chr.19: 14039878-14039878 on Build GRCh38
rs540624044 Chr.19: 14040036-14040036 on Build GRCh38
rs1005418235 Chr.19: 14040046-14040046 on Build GRCh38
rs767291532 Chr.19: 14039925-14039925 on Build GRCh38
rs774621762 Chr.19: 14039863-14039863 on Build GRCh38
rs376795280 Chr.19: 14039946-14039946 on Build GRCh38
rs536704566 Chr.19: 14039906-14039906 on Build GRCh38
rs74824519 Chr.19: 14040047-14040047 on Build GRCh38
rs143233265 Chr.19: 14039898-14039898 on Build GRCh38
rs772485804 Chr.19: 14039950-14039950 on Build GRCh38
rs748668943 Chr.19: 14039871-14039871 on Build GRCh38
rs201794701 Chr.19: 14039897-14039897 on Build GRCh38

View Full Product Details