Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166382601-166382848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC1
Entrez Gene ID
51660
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L, CGI-129, MPYCD, dJ68L15.3
UniGene
Hs.172755
Gene Chromosome Location
Chr.6: 166364919-166383013 on Build GRCh38
Transcripts
NM_001270879.1 NM_016098.3 NR_073093.1

SNP Detail

SNP ID Chromosome Location
rs1049763930 Chr.6: 166382634-166382634 on Build GRCh38
rs73265159 Chr.6: 166382738-166382738 on Build GRCh38
rs1002593456 Chr.6: 166382643-166382643 on Build GRCh38
rs776472438 Chr.6: 166382793-166382793 on Build GRCh38
rs938034650 Chr.6: 166382810-166382810 on Build GRCh38
rs745351305 Chr.6: 166382657-166382657 on Build GRCh38
rs781449430 Chr.6: 166382760-166382760 on Build GRCh38
rs950804669 Chr.6: 166382681-166382681 on Build GRCh38
rs777850458 Chr.6: 166382771-166382772 on Build GRCh38
rs374420236 Chr.6: 166382618-166382618 on Build GRCh38
rs550593206 Chr.6: 166382716-166382716 on Build GRCh38
rs202222055 Chr.6: 166382789-166382789 on Build GRCh38
rs944497057 Chr.6: 166382731-166382731 on Build GRCh38
rs528941946 Chr.6: 166382673-166382673 on Build GRCh38
rs566742239 Chr.6: 166382771-166382771 on Build GRCh38
rs377468342 Chr.6: 166382679-166382679 on Build GRCh38
rs1024186473 Chr.6: 166382656-166382656 on Build GRCh38
rs373243456 Chr.6: 166382765-166382765 on Build GRCh38
rs925845930 Chr.6: 166382676-166382676 on Build GRCh38
rs749057502 Chr.6: 166382762-166382762 on Build GRCh38
rs747983025 Chr.6: 166382774-166382774 on Build GRCh38
rs934978925 Chr.6: 166382780-166382780 on Build GRCh38
rs768378219 Chr.6: 166382772-166382772 on Build GRCh38
rs752740929 Chr.6: 166382644-166382644 on Build GRCh38
rs910302834 Chr.6: 166382722-166382722 on Build GRCh38
rs9459659 Chr.6: 166382754-166382754 on Build GRCh38
rs1049335304 Chr.6: 166382745-166382745 on Build GRCh38
rs542782757 Chr.6: 166382636-166382636 on Build GRCh38
rs1014098927 Chr.6: 166382654-166382654 on Build GRCh38
rs960747719 Chr.6: 166382651-166382651 on Build GRCh38
rs764728090 Chr.6: 166382819-166382819 on Build GRCh38
rs759949701 Chr.6: 166382794-166382794 on Build GRCh38
rs763301295 Chr.6: 166382809-166382809 on Build GRCh38
rs562340889 Chr.6: 166382734-166382734 on Build GRCh38
rs760329969 Chr.6: 166382784-166382784 on Build GRCh38
rs919454872 Chr.6: 166382737-166382737 on Build GRCh38
rs561473903 Chr.6: 166382650-166382650 on Build GRCh38
rs551398633 Chr.6: 166382755-166382755 on Build GRCh38
rs751623493 Chr.6: 166382822-166382822 on Build GRCh38
rs375956827 Chr.6: 166382801-166382801 on Build GRCh38
rs957072301 Chr.6: 166382709-166382709 on Build GRCh38
rs977396023 Chr.6: 166382669-166382669 on Build GRCh38
rs765478642 Chr.6: 166382796-166382796 on Build GRCh38

View Full Product Details