Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135010366-135010637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER3
Entrez Gene ID
347732
Gene Name
cation channel sperm associated 3
Gene Aliases
CACRC
UniGene
Hs.631804
Gene Chromosome Location
Chr.5: 134967906-135011707 on Build GRCh38
Transcripts
NM_178019.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6101926 c.1051A>T p.I351F Chr.5: 135010487-135010487 on Build GRCh38
6836566 c.1085C>T p.T362I Chr.5: 135010521-135010521 on Build GRCh38
448609 c.1034G>C p.S345T Chr.5: 135010470-135010470 on Build GRCh38
3138326 c.1066C>T p.L356L Chr.5: 135010502-135010502 on Build GRCh38
6202361 c.937-3T>A p.? Chr.5: 135010370-135010370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758367409 Chr.5: 135010603-135010603 on Build GRCh38
rs753181107 Chr.5: 135010539-135010539 on Build GRCh38
rs372406367 Chr.5: 135010570-135010570 on Build GRCh38
rs775607943 Chr.5: 135010538-135010538 on Build GRCh38
rs367634072 Chr.5: 135010484-135010484 on Build GRCh38
rs906601461 Chr.5: 135010502-135010502 on Build GRCh38
rs377081113 Chr.5: 135010578-135010578 on Build GRCh38
rs943249469 Chr.5: 135010467-135010467 on Build GRCh38
rs749967219 Chr.5: 135010469-135010469 on Build GRCh38
rs761386655 Chr.5: 135010454-135010454 on Build GRCh38
rs775375224 Chr.5: 135010431-135010431 on Build GRCh38
rs987159252 Chr.5: 135010607-135010607 on Build GRCh38
rs902540597 Chr.5: 135010589-135010589 on Build GRCh38
rs140770176 Chr.5: 135010483-135010483 on Build GRCh38
rs745967538 Chr.5: 135010520-135010520 on Build GRCh38
rs781691005 Chr.5: 135010518-135010518 on Build GRCh38
rs1011124053 Chr.5: 135010542-135010542 on Build GRCh38
rs772690944 Chr.5: 135010581-135010581 on Build GRCh38
rs17167765 Chr.5: 135010492-135010492 on Build GRCh38
rs771059196 Chr.5: 135010509-135010509 on Build GRCh38
rs877812 Chr.5: 135010614-135010614 on Build GRCh38
rs759476638 Chr.5: 135010399-135010399 on Build GRCh38
rs149182913 Chr.5: 135010511-135010511 on Build GRCh38
rs774022138 Chr.5: 135010573-135010573 on Build GRCh38
rs776860637 Chr.5: 135010397-135010397 on Build GRCh38
rs767179656 Chr.5: 135010580-135010580 on Build GRCh38
rs769830975 Chr.5: 135010523-135010523 on Build GRCh38
rs201141690 Chr.5: 135010499-135010499 on Build GRCh38
rs192161684 Chr.5: 135010613-135010613 on Build GRCh38
rs867207499 Chr.5: 135010476-135010476 on Build GRCh38
rs368429080 Chr.5: 135010541-135010541 on Build GRCh38
rs939382725 Chr.5: 135010506-135010506 on Build GRCh38
rs148358697 Chr.5: 135010477-135010477 on Build GRCh38
rs143349199 Chr.5: 135010447-135010447 on Build GRCh38
rs769883784 Chr.5: 135010427-135010427 on Build GRCh38
rs145588144 Chr.5: 135010485-135010485 on Build GRCh38
rs370122431 Chr.5: 135010497-135010497 on Build GRCh38
rs750244192 Chr.5: 135010585-135010585 on Build GRCh38
rs766192166 Chr.5: 135010396-135010396 on Build GRCh38
rs147914352 Chr.5: 135010445-135010445 on Build GRCh38
rs954601758 Chr.5: 135010606-135010606 on Build GRCh38
rs373800376 Chr.5: 135010457-135010457 on Build GRCh38
rs560060226 Chr.5: 135010574-135010574 on Build GRCh38

View Full Product Details