Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31424310-31424583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
UniGene
Hs.97997
Gene Chromosome Location
Chr.5: 31400494-31532175 on Build GRCh38
Transcripts
NM_001100412.1 NM_013235.4 XM_005248291.3 XM_005248292.3 XM_005248293.3 XM_005248294.3 XM_011514033.2 XM_017009399.1 XM_017009400.1 XM_017009401.1 XM_017009402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274875 c.3223C>T p.R1075C Chr.5: 31424465-31424465 on Build GRCh38
1067217 c.3239A>T p.N1080I Chr.5: 31424449-31424449 on Build GRCh38
6853407 c.3247C>T p.L1083F Chr.5: 31424441-31424441 on Build GRCh38
6314937 c.3225C>A p.R1075R Chr.5: 31424463-31424463 on Build GRCh38
5888849 c.3217-4C>T p.? Chr.5: 31424475-31424475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943915854 Chr.5: 31424482-31424482 on Build GRCh38
rs886778411 Chr.5: 31424558-31424558 on Build GRCh38
rs569922833 Chr.5: 31424518-31424518 on Build GRCh38
rs922017962 Chr.5: 31424361-31424361 on Build GRCh38
rs773556534 Chr.5: 31424428-31424429 on Build GRCh38
rs946918895 Chr.5: 31424486-31424486 on Build GRCh38
rs376727217 Chr.5: 31424465-31424465 on Build GRCh38
rs530099898 Chr.5: 31424414-31424414 on Build GRCh38
rs1057091184 Chr.5: 31424380-31424403 on Build GRCh38
rs755248297 Chr.5: 31424397-31424397 on Build GRCh38
rs773151368 Chr.5: 31424462-31424462 on Build GRCh38
rs552491336 Chr.5: 31424559-31424559 on Build GRCh38
rs767223074 Chr.5: 31424503-31424503 on Build GRCh38
rs995158342 Chr.5: 31424515-31424515 on Build GRCh38
rs750928345 Chr.5: 31424384-31424384 on Build GRCh38
rs781514950 Chr.5: 31424362-31424362 on Build GRCh38
rs373030516 Chr.5: 31424391-31424391 on Build GRCh38
rs968712619 Chr.5: 31424339-31424339 on Build GRCh38
rs200942350 Chr.5: 31424416-31424416 on Build GRCh38
rs769791099 Chr.5: 31424449-31424449 on Build GRCh38
rs979174741 Chr.5: 31424344-31424344 on Build GRCh38
rs927967183 Chr.5: 31424411-31424411 on Build GRCh38
rs377448843 Chr.5: 31424517-31424517 on Build GRCh38
rs532516956 Chr.5: 31424535-31424535 on Build GRCh38
rs988186391 Chr.5: 31424453-31424453 on Build GRCh38
rs867594081 Chr.5: 31424481-31424481 on Build GRCh38
rs765709467 Chr.5: 31424381-31424381 on Build GRCh38
rs934252066 Chr.5: 31424527-31424527 on Build GRCh38
rs561372899 Chr.5: 31424393-31424393 on Build GRCh38
rs1001264377 Chr.5: 31424544-31424544 on Build GRCh38
rs373877090 Chr.5: 31424499-31424499 on Build GRCh38
rs370032474 Chr.5: 31424497-31424497 on Build GRCh38
rs749231991 Chr.5: 31424463-31424463 on Build GRCh38
rs369580657 Chr.5: 31424387-31424387 on Build GRCh38

View Full Product Details