Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3013621-3013888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
UniGene
Hs.32959
Gene Chromosome Location
Chr.4: 2963504-3040760 on Build GRCh38
Transcripts
NM_001004056.1 NM_001004057.1 NM_005307.2 NM_182982.2 XM_005247962.3 XM_006713880.3 XM_011513447.2 XM_011513448.2 XM_011513449.2 XM_011513450.2 XM_011513451.2 XM_011513452.2 XM_011513453.2 XM_011513454.2 XM_011513455.2 XM_011513456.2 XM_011513457.2 XM_017008052.1 XM_017008053.1 XM_017008054.1 XM_017008055.1 XM_017008056.1 XM_017008057.1 XM_017008058.1 XM_017008059.1 XM_017008060.1 XM_017008061.1 XM_017008062.1 XM_017008063.1 XM_017008064.1 XM_017008065.1 XM_017008066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380907 c.721A>G p.K241E Chr.4: 3013808-3013808 on Build GRCh38
4765018 c.656_657insAA p.R222fs*3 Chr.4: 3013743-3013744 on Build GRCh38
4332209 c.655C>A p.Q219K Chr.4: 3013742-3013742 on Build GRCh38
4124316 c.697C>T p.L233L Chr.4: 3013784-3013784 on Build GRCh38
1179565 c.655_656insA p.R222fs*8 Chr.4: 3013742-3013743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778027536 Chr.4: 3013793-3013793 on Build GRCh38
rs757898806 Chr.4: 3013749-3013749 on Build GRCh38
rs769163351 Chr.4: 3013693-3013693 on Build GRCh38
rs372439107 Chr.4: 3013856-3013856 on Build GRCh38
rs763819143 Chr.4: 3013734-3013734 on Build GRCh38
rs770112448 Chr.4: 3013702-3013702 on Build GRCh38
rs771118594 Chr.4: 3013809-3013809 on Build GRCh38
rs903972349 Chr.4: 3013723-3013723 on Build GRCh38
rs774297658 Chr.4: 3013714-3013714 on Build GRCh38
rs749991657 Chr.4: 3013744-3013744 on Build GRCh38
rs931491157 Chr.4: 3013860-3013860 on Build GRCh38
rs1033686924 Chr.4: 3013759-3013759 on Build GRCh38
rs1019611230 Chr.4: 3013794-3013794 on Build GRCh38
rs753629667 Chr.4: 3013739-3013739 on Build GRCh38
rs113286884 Chr.4: 3013699-3013699 on Build GRCh38
rs773152983 Chr.4: 3013726-3013726 on Build GRCh38
rs745320025 Chr.4: 3013684-3013684 on Build GRCh38
rs562227299 Chr.4: 3013700-3013700 on Build GRCh38
rs763102146 Chr.4: 3013707-3013707 on Build GRCh38
rs368284634 Chr.4: 3013784-3013784 on Build GRCh38
rs199873516 Chr.4: 3013797-3013797 on Build GRCh38
rs773527943 Chr.4: 3013706-3013706 on Build GRCh38
rs778319834 Chr.4: 3013680-3013680 on Build GRCh38
rs760726349 Chr.4: 3013861-3013861 on Build GRCh38
rs200846796 Chr.4: 3013772-3013772 on Build GRCh38
rs777526337 Chr.4: 3013664-3013664 on Build GRCh38
rs1051177201 Chr.4: 3013687-3013687 on Build GRCh38
rs762202589 Chr.4: 3013743-3013743 on Build GRCh38
rs759677405 Chr.4: 3013728-3013728 on Build GRCh38
rs992087865 Chr.4: 3013801-3013801 on Build GRCh38
rs984661786 Chr.4: 3013847-3013847 on Build GRCh38
rs900048609 Chr.4: 3013649-3013649 on Build GRCh38
rs762319249 Chr.4: 3013665-3013666 on Build GRCh38
rs748628207 Chr.4: 3013780-3013780 on Build GRCh38
rs753411169 Chr.4: 3013675-3013675 on Build GRCh38
rs756866243 Chr.4: 3013742-3013742 on Build GRCh38
rs755973293 Chr.4: 3013647-3013647 on Build GRCh38
rs764884675 Chr.4: 3013743-3013743 on Build GRCh38
rs774631574 Chr.4: 3013813-3013813 on Build GRCh38
rs572038787 Chr.4: 3013694-3013694 on Build GRCh38
rs771141508 Chr.4: 3013712-3013712 on Build GRCh38
rs375862689 Chr.4: 3013751-3013752 on Build GRCh38
rs775540841 Chr.4: 3013731-3013731 on Build GRCh38
rs1140085 Chr.4: 3013826-3013826 on Build GRCh38
rs1009716084 Chr.4: 3013698-3013698 on Build GRCh38
rs35824641 Chr.4: 3013730-3013730 on Build GRCh38
rs556199273 Chr.4: 3013742-3013743 on Build GRCh38
rs745989304 Chr.4: 3013815-3013815 on Build GRCh38

View Full Product Details