Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44506174-44506433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCB
Entrez Gene ID
84075
Gene Name
fibrous sheath CABYR binding protein
Gene Aliases
C14orf155
UniGene
Hs.307086
Gene Chromosome Location
Chr.14: 44504149-44507296 on Build GRCh38
Transcripts
NM_032135.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1369822 c.632A>T p.K211I Chr.14: 44506356-44506356 on Build GRCh38
6305150 c.560C>A p.S187* Chr.14: 44506428-44506428 on Build GRCh38
3734924 c.736G>A p.G246S Chr.14: 44506252-44506252 on Build GRCh38
6028313 c.671_672insA p.G225fs*9 Chr.14: 44506316-44506317 on Build GRCh38
2197142 c.700C>T p.L234F Chr.14: 44506288-44506288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762872354 Chr.14: 44506341-44506341 on Build GRCh38
rs774769264 Chr.14: 44506316-44506316 on Build GRCh38
rs76172359 Chr.14: 44506316-44506316 on Build GRCh38
rs772460438 Chr.14: 44506384-44506387 on Build GRCh38
rs374505767 Chr.14: 44506302-44506302 on Build GRCh38
rs3809430 Chr.14: 44506307-44506307 on Build GRCh38
rs746185851 Chr.14: 44506379-44506380 on Build GRCh38
rs558630150 Chr.14: 44506311-44506311 on Build GRCh38
rs746865726 Chr.14: 44506330-44506330 on Build GRCh38
rs372393712 Chr.14: 44506270-44506270 on Build GRCh38
rs774628287 Chr.14: 44506351-44506351 on Build GRCh38
rs199704834 Chr.14: 44506408-44506408 on Build GRCh38
rs775626824 Chr.14: 44506387-44506388 on Build GRCh38
rs745606014 Chr.14: 44506222-44506222 on Build GRCh38
rs762376046 Chr.14: 44506308-44506308 on Build GRCh38
rs774292188 Chr.14: 44506217-44506217 on Build GRCh38
rs3809429 Chr.14: 44506403-44506403 on Build GRCh38
rs374207258 Chr.14: 44506247-44506247 on Build GRCh38
rs770730262 Chr.14: 44506208-44506208 on Build GRCh38
rs866589510 Chr.14: 44506384-44506385 on Build GRCh38
rs752963679 Chr.14: 44506368-44506368 on Build GRCh38
rs753962867 Chr.14: 44506251-44506251 on Build GRCh38
rs748866831 Chr.14: 44506206-44506206 on Build GRCh38
rs771860041 Chr.14: 44506227-44506227 on Build GRCh38
rs368901910 Chr.14: 44506256-44506256 on Build GRCh38
rs45478391 Chr.14: 44506308-44506308 on Build GRCh38
rs760903043 Chr.14: 44506401-44506401 on Build GRCh38
rs768178319 Chr.14: 44506364-44506364 on Build GRCh38
rs772056085 Chr.14: 44506324-44506324 on Build GRCh38
rs758303849 Chr.14: 44506397-44506397 on Build GRCh38
rs775332427 Chr.14: 44506231-44506231 on Build GRCh38
rs748817081 Chr.14: 44506265-44506265 on Build GRCh38
rs544184497 Chr.14: 44506370-44506370 on Build GRCh38
rs944930818 Chr.14: 44506331-44506331 on Build GRCh38
rs576897354 Chr.14: 44506349-44506349 on Build GRCh38
rs753534693 Chr.14: 44506379-44506379 on Build GRCh38
rs202120862 Chr.14: 44506393-44506393 on Build GRCh38
rs140155770 Chr.14: 44506354-44506354 on Build GRCh38
rs199558988 Chr.14: 44506267-44506267 on Build GRCh38
rs760094503 Chr.14: 44506361-44506361 on Build GRCh38
rs773138383 Chr.14: 44506240-44506240 on Build GRCh38
rs777325266 Chr.14: 44506234-44506235 on Build GRCh38
rs897982395 Chr.14: 44506375-44506375 on Build GRCh38
rs529179674 Chr.14: 44506363-44506363 on Build GRCh38
rs758692792 Chr.14: 44506377-44506377 on Build GRCh38
rs780002065 Chr.14: 44506402-44506402 on Build GRCh38
rs771438058 Chr.14: 44506269-44506270 on Build GRCh38
rs755606474 Chr.14: 44506253-44506253 on Build GRCh38
rs376229577 Chr.14: 44506312-44506312 on Build GRCh38
rs796111835 Chr.14: 44506218-44506218 on Build GRCh38
rs1036602003 Chr.14: 44506353-44506353 on Build GRCh38
rs778382650 Chr.14: 44506390-44506390 on Build GRCh38
rs756927626 Chr.14: 44506381-44506381 on Build GRCh38
rs748456747 Chr.14: 44506332-44506332 on Build GRCh38
rs146549008 Chr.14: 44506337-44506337 on Build GRCh38
rs554830800 Chr.14: 44506237-44506237 on Build GRCh38
rs777512025 Chr.14: 44506284-44506284 on Build GRCh38
rs755690567 Chr.14: 44506283-44506283 on Build GRCh38
rs775248225 Chr.14: 44506328-44506328 on Build GRCh38
rs35849240 Chr.14: 44506236-44506236 on Build GRCh38

View Full Product Details