Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8863716-8863943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO1
Entrez Gene ID
2023
Gene Name
enolase 1
Gene Aliases
ENO1L1, HEL-S-17, MPB1, NNE, PPH
UniGene
Hs.517145
Gene Chromosome Location
Chr.1: 8861000-8879092 on Build GRCh38
Transcripts
NM_001201483.1 NM_001428.3 XM_006710433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4749642 c.1064A>G p.Q355R Chr.1: 8863894-8863894 on Build GRCh38
4882657 c.1038G>C p.Q346H Chr.1: 8863920-8863920 on Build GRCh38
4456670 c.1018C>T p.L340F Chr.1: 8863940-8863940 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs921276137 Chr.1: 8863846-8863846 on Build GRCh38
rs771892272 Chr.1: 8863882-8863882 on Build GRCh38
rs375185398 Chr.1: 8863792-8863792 on Build GRCh38
rs759414399 Chr.1: 8863895-8863895 on Build GRCh38
rs536666214 Chr.1: 8863851-8863851 on Build GRCh38
rs750467514 Chr.1: 8863910-8863910 on Build GRCh38
rs981732152 Chr.1: 8863805-8863805 on Build GRCh38
rs28999093 Chr.1: 8863852-8863852 on Build GRCh38
rs765810181 Chr.1: 8863890-8863890 on Build GRCh38
rs878870368 Chr.1: 8863888-8863889 on Build GRCh38
rs756568186 Chr.1: 8863869-8863869 on Build GRCh38
rs769087479 Chr.1: 8863873-8863873 on Build GRCh38
rs761998408 Chr.1: 8863907-8863907 on Build GRCh38
rs367986914 Chr.1: 8863911-8863911 on Build GRCh38
rs767786982 Chr.1: 8863908-8863908 on Build GRCh38
rs1023030986 Chr.1: 8863786-8863786 on Build GRCh38
rs764967196 Chr.1: 8863904-8863904 on Build GRCh38
rs772576829 Chr.1: 8863885-8863885 on Build GRCh38
rs747764179 Chr.1: 8863877-8863877 on Build GRCh38
rs757497928 Chr.1: 8863860-8863860 on Build GRCh38
rs201636531 Chr.1: 8863894-8863894 on Build GRCh38
rs992637435 Chr.1: 8863781-8863781 on Build GRCh38
rs28999094 Chr.1: 8863782-8863782 on Build GRCh38
rs756310850 Chr.1: 8863787-8863793 on Build GRCh38
rs746788399 Chr.1: 8863796-8863796 on Build GRCh38
rs751727605 Chr.1: 8863858-8863858 on Build GRCh38
rs570459529 Chr.1: 8863868-8863868 on Build GRCh38
rs951785102 Chr.1: 8863777-8863777 on Build GRCh38
rs548503758 Chr.1: 8863746-8863746 on Build GRCh38
rs375359255 Chr.1: 8863872-8863872 on Build GRCh38
rs781215262 Chr.1: 8863861-8863861 on Build GRCh38
rs774878060 Chr.1: 8863874-8863874 on Build GRCh38
rs752525268 Chr.1: 8863905-8863905 on Build GRCh38
rs566364027 Chr.1: 8863839-8863839 on Build GRCh38
rs971089608 Chr.1: 8863790-8863790 on Build GRCh38
rs776573770 Chr.1: 8863891-8863891 on Build GRCh38
rs780680746 Chr.1: 8863870-8863870 on Build GRCh38
rs1027132323 Chr.1: 8863766-8863766 on Build GRCh38
rs1026594414 Chr.1: 8863743-8863743 on Build GRCh38
rs754321568 Chr.1: 8863922-8863922 on Build GRCh38
rs755573568 Chr.1: 8863923-8863923 on Build GRCh38
rs760298320 Chr.1: 8863888-8863888 on Build GRCh38

View Full Product Details