Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52800719-52800954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373908 c.367G>A p.E123K Chr.5: 52800761-52800761 on Build GRCh38
4347979 c.371G>C p.R124P Chr.5: 52800765-52800765 on Build GRCh38
1287076 c.534T>A p.H178Q Chr.5: 52800928-52800928 on Build GRCh38
5137272 c.331A>T p.T111S Chr.5: 52800725-52800725 on Build GRCh38
3616898 c.330C>T p.F110F Chr.5: 52800724-52800724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771365583 Chr.5: 52800793-52800793 on Build GRCh38
rs746188540 Chr.5: 52800812-52800812 on Build GRCh38
rs548469886 Chr.5: 52800752-52800752 on Build GRCh38
rs1024183492 Chr.5: 52800833-52800833 on Build GRCh38
rs772615662 Chr.5: 52800839-52800839 on Build GRCh38
rs139161716 Chr.5: 52800769-52800769 on Build GRCh38
rs991725772 Chr.5: 52800807-52800807 on Build GRCh38
rs770338598 Chr.5: 52800813-52800813 on Build GRCh38
rs761560784 Chr.5: 52800824-52800824 on Build GRCh38
rs549701701 Chr.5: 52800918-52800918 on Build GRCh38
rs200636575 Chr.5: 52800779-52800779 on Build GRCh38
rs777861421 Chr.5: 52800902-52800902 on Build GRCh38
rs753877060 Chr.5: 52800878-52800878 on Build GRCh38
rs958659616 Chr.5: 52800773-52800773 on Build GRCh38
rs771667455 Chr.5: 52800836-52800836 on Build GRCh38
rs750307879 Chr.5: 52800763-52800763 on Build GRCh38
rs971203469 Chr.5: 52800866-52800866 on Build GRCh38
rs774088225 Chr.5: 52800818-52800818 on Build GRCh38
rs761392908 Chr.5: 52800749-52800749 on Build GRCh38
rs144148189 Chr.5: 52800785-52800785 on Build GRCh38
rs143871783 Chr.5: 52800811-52800812 on Build GRCh38
rs752854138 Chr.5: 52800896-52800896 on Build GRCh38
rs202063626 Chr.5: 52800788-52800788 on Build GRCh38
rs779106178 Chr.5: 52800778-52800778 on Build GRCh38
rs754805559 Chr.5: 52800774-52800774 on Build GRCh38
rs758654730 Chr.5: 52800900-52800900 on Build GRCh38
rs781748090 Chr.5: 52800805-52800805 on Build GRCh38
rs776373330 Chr.5: 52800787-52800788 on Build GRCh38
rs774047496 Chr.5: 52800739-52800739 on Build GRCh38
rs368443275 Chr.5: 52800789-52800789 on Build GRCh38
rs753550497 Chr.5: 52800772-52800772 on Build GRCh38
rs766144124 Chr.5: 52800770-52800770 on Build GRCh38
rs751570929 Chr.5: 52800915-52800915 on Build GRCh38
rs765033407 Chr.5: 52800892-52800892 on Build GRCh38
rs35160817 Chr.5: 52800841-52800841 on Build GRCh38
rs747511590 Chr.5: 52800791-52800791 on Build GRCh38
rs35749418 Chr.5: 52800798-52800798 on Build GRCh38
rs760277990 Chr.5: 52800852-52800852 on Build GRCh38
rs759316671 Chr.5: 52800881-52800881 on Build GRCh38
rs766192469 Chr.5: 52800874-52800874 on Build GRCh38
rs750569887 Chr.5: 52800827-52800827 on Build GRCh38

View Full Product Details