Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62361363-62361594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
UniGene
Hs.558351
Gene Chromosome Location
Chr.5: 62306162-62387184 on Build GRCh38
Transcripts
NM_001098511.2 NM_001243952.1 NM_001243953.1 NM_004520.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210926 c.942A>C p.L314F Chr.5: 62361525-62361525 on Build GRCh38
6210925 c.1023A>C p.L341F Chr.5: 62361525-62361525 on Build GRCh38
4590350 c.992A>G p.N331S Chr.5: 62361494-62361494 on Build GRCh38
4590351 c.911A>G p.N304S Chr.5: 62361494-62361494 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563961990 Chr.5: 62361454-62361454 on Build GRCh38
rs752464946 Chr.5: 62361480-62361480 on Build GRCh38
rs765866312 Chr.5: 62361425-62361425 on Build GRCh38
rs247265 Chr.5: 62361401-62361401 on Build GRCh38
rs758143705 Chr.5: 62361526-62361526 on Build GRCh38
rs781390897 Chr.5: 62361544-62361544 on Build GRCh38
rs962980768 Chr.5: 62361436-62361436 on Build GRCh38
rs551258913 Chr.5: 62361405-62361405 on Build GRCh38
rs760232847 Chr.5: 62361421-62361421 on Build GRCh38
rs751983502 Chr.5: 62361498-62361498 on Build GRCh38
rs781229638 Chr.5: 62361535-62361536 on Build GRCh38
rs751795324 Chr.5: 62361535-62361535 on Build GRCh38
rs776587016 Chr.5: 62361435-62361435 on Build GRCh38
rs973160246 Chr.5: 62361444-62361444 on Build GRCh38
rs925330883 Chr.5: 62361450-62361450 on Build GRCh38
rs200648499 Chr.5: 62361538-62361538 on Build GRCh38
rs765109308 Chr.5: 62361468-62361468 on Build GRCh38
rs776555508 Chr.5: 62361404-62361404 on Build GRCh38
rs764238744 Chr.5: 62361533-62361533 on Build GRCh38
rs958348972 Chr.5: 62361494-62361494 on Build GRCh38
rs759447626 Chr.5: 62361437-62361437 on Build GRCh38
rs247266 Chr.5: 62361550-62361550 on Build GRCh38

View Full Product Details