Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56992379-56992614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND6
Entrez Gene ID
221336
Gene Name
BEN domain containing 6
Gene Aliases
C6orf65
UniGene
Hs.582993
Gene Chromosome Location
Chr.6: 56954975-57027652 on Build GRCh38
Transcripts
NM_001318539.1 NM_152731.2 XM_011514346.2 XM_017010406.1 XM_017010407.1 XM_017010408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6107294 c.253C>A p.R85R Chr.6: 56992510-56992510 on Build GRCh38
5200725 c.288C>A p.V96V Chr.6: 56992545-56992545 on Build GRCh38
6107290 c.283T>A p.L95M Chr.6: 56992540-56992540 on Build GRCh38
3830898 c.250A>C p.T84P Chr.6: 56992507-56992507 on Build GRCh38
4483176 c.267C>T p.S89S Chr.6: 56992524-56992524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79968708 Chr.6: 56992516-56992516 on Build GRCh38
rs969490420 Chr.6: 56992518-56992518 on Build GRCh38
rs761993055 Chr.6: 56992488-56992488 on Build GRCh38
rs765191178 Chr.6: 56992492-56992492 on Build GRCh38
rs1011420338 Chr.6: 56992567-56992567 on Build GRCh38
rs928555916 Chr.6: 56992525-56992525 on Build GRCh38
rs769056468 Chr.6: 56992479-56992479 on Build GRCh38
rs377361378 Chr.6: 56992470-56992470 on Build GRCh38
rs746259544 Chr.6: 56992487-56992487 on Build GRCh38
rs113465738 Chr.6: 56992503-56992503 on Build GRCh38
rs758819129 Chr.6: 56992412-56992412 on Build GRCh38
rs775360572 Chr.6: 56992476-56992476 on Build GRCh38
rs892934040 Chr.6: 56992546-56992546 on Build GRCh38
rs1045833040 Chr.6: 56992512-56992512 on Build GRCh38
rs755317583 Chr.6: 56992511-56992511 on Build GRCh38
rs752357616 Chr.6: 56992425-56992425 on Build GRCh38
rs750958672 Chr.6: 56992498-56992498 on Build GRCh38
rs780343975 Chr.6: 56992423-56992423 on Build GRCh38
rs377420280 Chr.6: 56992520-56992521 on Build GRCh38
rs770321168 Chr.6: 56992465-56992465 on Build GRCh38
rs770094003 Chr.6: 56992531-56992532 on Build GRCh38
rs763306817 Chr.6: 56992499-56992499 on Build GRCh38
rs756814276 Chr.6: 56992532-56992532 on Build GRCh38
rs750863070 Chr.6: 56992407-56992407 on Build GRCh38
rs1052954048 Chr.6: 56992531-56992531 on Build GRCh38
rs371013253 Chr.6: 56992471-56992471 on Build GRCh38
rs763778967 Chr.6: 56992521-56992521 on Build GRCh38
rs778228205 Chr.6: 56992564-56992564 on Build GRCh38
rs748881219 Chr.6: 56992461-56992461 on Build GRCh38
rs762196915 Chr.6: 56992510-56992510 on Build GRCh38
rs866381843 Chr.6: 56992435-56992435 on Build GRCh38
rs776968928 Chr.6: 56992483-56992483 on Build GRCh38
rs201522335 Chr.6: 56992437-56992437 on Build GRCh38
rs778944023 Chr.6: 56992468-56992468 on Build GRCh38
rs777394418 Chr.6: 56992449-56992449 on Build GRCh38
rs201589903 Chr.6: 56992526-56992526 on Build GRCh38
rs760544105 Chr.6: 56992477-56992477 on Build GRCh38
rs199677690 Chr.6: 56992485-56992485 on Build GRCh38

View Full Product Details