Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96840271-96840544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645478 c.433C>T p.R145C Chr.2: 96840508-96840508 on Build GRCh38
1023996 c.496G>A p.A166T Chr.2: 96840445-96840445 on Build GRCh38
4661804 c.513C>T p.D171D Chr.2: 96840428-96840428 on Build GRCh38
4759136 c.580A>C p.K194Q Chr.2: 96840276-96840276 on Build GRCh38
6600475 c.514G>A p.A172T Chr.2: 96840427-96840427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772252975 Chr.2: 96840297-96840297 on Build GRCh38
rs769361189 Chr.2: 96840334-96840334 on Build GRCh38
rs761537350 Chr.2: 96840434-96840434 on Build GRCh38
rs762652961 Chr.2: 96840357-96840357 on Build GRCh38
rs746554016 Chr.2: 96840394-96840395 on Build GRCh38
rs959026394 Chr.2: 96840470-96840470 on Build GRCh38
rs765786216 Chr.2: 96840445-96840445 on Build GRCh38
rs779606396 Chr.2: 96840390-96840390 on Build GRCh38
rs746828314 Chr.2: 96840306-96840306 on Build GRCh38
rs766666488 Chr.2: 96840439-96840439 on Build GRCh38
rs758370218 Chr.2: 96840448-96840448 on Build GRCh38
rs764258615 Chr.2: 96840463-96840463 on Build GRCh38
rs758624990 Chr.2: 96840415-96840415 on Build GRCh38
rs1049662274 Chr.2: 96840333-96840333 on Build GRCh38
rs777049792 Chr.2: 96840440-96840440 on Build GRCh38
rs377168591 Chr.2: 96840314-96840314 on Build GRCh38
rs984296295 Chr.2: 96840442-96840442 on Build GRCh38
rs777614482 Chr.2: 96840298-96840298 on Build GRCh38
rs997347390 Chr.2: 96840388-96840389 on Build GRCh38
rs781137462 Chr.2: 96840424-96840424 on Build GRCh38
rs747753720 Chr.2: 96840504-96840504 on Build GRCh38
rs1041146996 Chr.2: 96840367-96840367 on Build GRCh38
rs1007011539 Chr.2: 96840351-96840351 on Build GRCh38
rs766750293 Chr.2: 96840392-96840392 on Build GRCh38
rs776961850 Chr.2: 96840511-96840511 on Build GRCh38
rs750764531 Chr.2: 96840384-96840384 on Build GRCh38
rs758053404 Chr.2: 96840375-96840392 on Build GRCh38
rs752790639 Chr.2: 96840446-96840446 on Build GRCh38
rs752781835 Chr.2: 96840408-96840408 on Build GRCh38
rs534817117 Chr.2: 96840347-96840347 on Build GRCh38
rs371726283 Chr.2: 96840427-96840427 on Build GRCh38
rs1010715183 Chr.2: 96840372-96840372 on Build GRCh38
rs561812290 Chr.2: 96840478-96840478 on Build GRCh38
rs781121082 Chr.2: 96840475-96840475 on Build GRCh38
rs554610108 Chr.2: 96840423-96840423 on Build GRCh38
rs116410092 Chr.2: 96840481-96840481 on Build GRCh38
rs375475998 Chr.2: 96840431-96840431 on Build GRCh38
rs760026329 Chr.2: 96840444-96840444 on Build GRCh38
rs751675667 Chr.2: 96840469-96840469 on Build GRCh38
rs769936679 Chr.2: 96840507-96840507 on Build GRCh38
rs759934194 Chr.2: 96840516-96840516 on Build GRCh38
rs370284092 Chr.2: 96840502-96840502 on Build GRCh38
rs372189853 Chr.2: 96840385-96840385 on Build GRCh38
rs750012079 Chr.2: 96840347-96840347 on Build GRCh38
rs769908649 Chr.2: 96840426-96840426 on Build GRCh38
rs114122128 Chr.2: 96840473-96840473 on Build GRCh38
rs777898323 Chr.2: 96840422-96840422 on Build GRCh38
rs778695344 Chr.2: 96840304-96840304 on Build GRCh38
rs901699796 Chr.2: 96840387-96840387 on Build GRCh38
rs759102995 Chr.2: 96840425-96840425 on Build GRCh38
rs759558510 Chr.2: 96840317-96840317 on Build GRCh38
rs762406291 Chr.2: 96840428-96840428 on Build GRCh38
rs768240120 Chr.2: 96840433-96840433 on Build GRCh38
rs755483488 Chr.2: 96840399-96840399 on Build GRCh38
rs754288228 Chr.2: 96840395-96840395 on Build GRCh38
rs144783054 Chr.2: 96840503-96840503 on Build GRCh38
rs770871544 Chr.2: 96840313-96840313 on Build GRCh38
rs778963397 Chr.2: 96840400-96840400 on Build GRCh38
rs763698489 Chr.2: 96840381-96840381 on Build GRCh38
rs771773900 Chr.2: 96840508-96840508 on Build GRCh38
rs749267905 Chr.2: 96840497-96840497 on Build GRCh38
rs922249350 Chr.2: 96840487-96840487 on Build GRCh38
rs893622497 Chr.2: 96840366-96840366 on Build GRCh38
rs369279629 Chr.2: 96840491-96840491 on Build GRCh38

View Full Product Details