Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4969973-4970198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA2
Entrez Gene ID
23125
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
-
UniGene
Hs.632242
Gene Chromosome Location
Chr.17: 4967992-4988297 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2 NR_106925.1

Gene Symbol
MIR6865
Entrez Gene ID
102465522
Gene Name
microRNA 6865
Gene Aliases
hsa-mir-6865
UniGene
Gene Chromosome Location
Chr.17: 4970086-4970150 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2 NR_106925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7330734 c.3073C>G p.L1025V Chr.17: 4970024-4970024 on Build GRCh38
6538621 c.3075A>C p.A1025A Chr.17: 4970016-4970016 on Build GRCh38
1283918 c.3076T>G p.S1026A Chr.17: 4970015-4970015 on Build GRCh38
4486915 c.3119C>T p.P1040L Chr.17: 4970041-4970041 on Build GRCh38
7330733 c.3067C>G p.L1023V Chr.17: 4970024-4970024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369189393 Chr.17: 4970129-4970129 on Build GRCh38
rs777003581 Chr.17: 4970063-4970063 on Build GRCh38
rs756717758 Chr.17: 4970113-4970113 on Build GRCh38
rs777350901 Chr.17: 4970099-4970099 on Build GRCh38
rs370933374 Chr.17: 4970145-4970145 on Build GRCh38
rs913656035 Chr.17: 4970134-4970134 on Build GRCh38
rs1024865919 Chr.17: 4970052-4970052 on Build GRCh38
rs753494379 Chr.17: 4970108-4970108 on Build GRCh38
rs752611767 Chr.17: 4970144-4970145 on Build GRCh38
rs189574616 Chr.17: 4970078-4970078 on Build GRCh38
rs559152019 Chr.17: 4970076-4970076 on Build GRCh38
rs756494271 Chr.17: 4970155-4970155 on Build GRCh38
rs745435850 Chr.17: 4970132-4970132 on Build GRCh38
rs767356395 Chr.17: 4970143-4970144 on Build GRCh38
rs773134384 Chr.17: 4970172-4970172 on Build GRCh38
rs764409720 Chr.17: 4970062-4970062 on Build GRCh38
rs770103046 Chr.17: 4970164-4970164 on Build GRCh38
rs746394672 Chr.17: 4970041-4970041 on Build GRCh38
rs772430030 Chr.17: 4970143-4970143 on Build GRCh38
rs752428269 Chr.17: 4970093-4970093 on Build GRCh38
rs772437234 Chr.17: 4970042-4970042 on Build GRCh38
rs762145165 Chr.17: 4970157-4970157 on Build GRCh38
rs970663882 Chr.17: 4970054-4970054 on Build GRCh38
rs1047236116 Chr.17: 4970142-4970142 on Build GRCh38
rs755961880 Chr.17: 4970094-4970094 on Build GRCh38
rs780680742 Chr.17: 4970144-4970144 on Build GRCh38
rs377674300 Chr.17: 4970082-4970082 on Build GRCh38
rs989222017 Chr.17: 4970100-4970100 on Build GRCh38
rs966814795 Chr.17: 4970136-4970136 on Build GRCh38
rs374140630 Chr.17: 4970049-4970049 on Build GRCh38
rs769144513 Chr.17: 4970152-4970152 on Build GRCh38
rs761914563 Chr.17: 4970068-4970068 on Build GRCh38
rs765435073 Chr.17: 4970073-4970073 on Build GRCh38
rs753057836 Chr.17: 4970037-4970037 on Build GRCh38
rs761144635 Chr.17: 4970056-4970056 on Build GRCh38

View Full Product Details