Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48796375-48796648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT2
Entrez Gene ID
587
Gene Name
branched chain amino acid transaminase 2
Gene Aliases
BCAM, BCATM, BCT2, PP18
UniGene
Hs.512670
Gene Chromosome Location
Chr.19: 48795062-48811063 on Build GRCh38
Transcripts
NM_001164773.1 NM_001190.3 NM_001284325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1580562 c.1004T>C p.V335A Chr.19: 48796639-48796639 on Build GRCh38
1580563 c.997C>T p.R333W Chr.19: 48796646-48796646 on Build GRCh38
5390265 c.1107C>T p.I369I Chr.19: 48796461-48796461 on Build GRCh38
5856338 c.1046G>A p.R349Q Chr.19: 48796597-48796597 on Build GRCh38
146163 c.1065+2T>A p.? Chr.19: 48796576-48796576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748466142 Chr.19: 48796511-48796511 on Build GRCh38
rs763574920 Chr.19: 48796554-48796554 on Build GRCh38
rs913296741 Chr.19: 48796475-48796475 on Build GRCh38
rs138253411 Chr.19: 48796614-48796614 on Build GRCh38
rs201393521 Chr.19: 48796555-48796555 on Build GRCh38
rs879000788 Chr.19: 48796485-48796485 on Build GRCh38
rs755170050 Chr.19: 48796501-48796501 on Build GRCh38
rs763609699 Chr.19: 48796409-48796409 on Build GRCh38
rs774810496 Chr.19: 48796525-48796525 on Build GRCh38
rs560632911 Chr.19: 48796552-48796552 on Build GRCh38
rs369301424 Chr.19: 48796408-48796408 on Build GRCh38
rs779570956 Chr.19: 48796622-48796622 on Build GRCh38
rs376034825 Chr.19: 48796407-48796407 on Build GRCh38
rs980319890 Chr.19: 48796431-48796431 on Build GRCh38
rs772311929 Chr.19: 48796630-48796630 on Build GRCh38
rs571191767 Chr.19: 48796629-48796629 on Build GRCh38
rs769745841 Chr.19: 48796512-48796512 on Build GRCh38
rs372960566 Chr.19: 48796519-48796519 on Build GRCh38
rs749404872 Chr.19: 48796612-48796612 on Build GRCh38
rs757236406 Chr.19: 48796462-48796462 on Build GRCh38
rs771397384 Chr.19: 48796423-48796423 on Build GRCh38
rs140856269 Chr.19: 48796448-48796448 on Build GRCh38
rs771599764 Chr.19: 48796520-48796520 on Build GRCh38
rs530143373 Chr.19: 48796518-48796518 on Build GRCh38
rs781275517 Chr.19: 48796507-48796507 on Build GRCh38
rs759835559 Chr.19: 48796548-48796548 on Build GRCh38
rs920381904 Chr.19: 48796398-48796398 on Build GRCh38
rs766250634 Chr.19: 48796573-48796573 on Build GRCh38
rs1132361 Chr.19: 48796494-48796494 on Build GRCh38
rs759822965 Chr.19: 48796404-48796405 on Build GRCh38
rs765578328 Chr.19: 48796538-48796539 on Build GRCh38
rs748189386 Chr.19: 48796401-48796401 on Build GRCh38
rs199999090 Chr.19: 48796457-48796457 on Build GRCh38
rs562507811 Chr.19: 48796474-48796474 on Build GRCh38
rs777190327 Chr.19: 48796400-48796418 on Build GRCh38
rs752836872 Chr.19: 48796597-48796597 on Build GRCh38
rs750239266 Chr.19: 48796493-48796493 on Build GRCh38
rs760435320 Chr.19: 48796553-48796553 on Build GRCh38
rs759145627 Chr.19: 48796587-48796587 on Build GRCh38
rs146438052 Chr.19: 48796442-48796442 on Build GRCh38
rs564868026 Chr.19: 48796418-48796418 on Build GRCh38
rs760111599 Chr.19: 48796449-48796449 on Build GRCh38
rs150116784 Chr.19: 48796456-48796456 on Build GRCh38
rs771693294 Chr.19: 48796549-48796549 on Build GRCh38
rs35829476 Chr.19: 48796570-48796570 on Build GRCh38
rs756442340 Chr.19: 48796604-48796604 on Build GRCh38
rs761687961 Chr.19: 48796561-48796561 on Build GRCh38
rs993016942 Chr.19: 48796468-48796468 on Build GRCh38
rs201944449 Chr.19: 48796571-48796571 on Build GRCh38
rs767101452 Chr.19: 48796445-48796445 on Build GRCh38
rs765172319 Chr.19: 48796568-48796568 on Build GRCh38

View Full Product Details