Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6728088-6728347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS7A
Entrez Gene ID
50813
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
CSN7, CSN7A, SGN7a
UniGene
Hs.530823
Gene Chromosome Location
Chr.12: 6723984-6731875 on Build GRCh38
Transcripts
NM_001164093.1 NM_001164094.1 NM_001164095.2 NM_016319.3 XM_005253694.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4396385 c.254T>G p.L85R Chr.12: 6728238-6728238 on Build GRCh38
942831 c.290G>A p.R97Q Chr.12: 6728274-6728274 on Build GRCh38
416100 c.256C>G p.P86A Chr.12: 6728240-6728240 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182144203 Chr.12: 6728212-6728212 on Build GRCh38
rs778782482 Chr.12: 6728191-6728191 on Build GRCh38
rs760743519 Chr.12: 6728263-6728263 on Build GRCh38
rs147088395 Chr.12: 6728232-6728232 on Build GRCh38
rs758156967 Chr.12: 6728203-6728203 on Build GRCh38
rs1050475460 Chr.12: 6728137-6728137 on Build GRCh38
rs750965855 Chr.12: 6728164-6728164 on Build GRCh38
rs1023271179 Chr.12: 6728231-6728231 on Build GRCh38
rs1014678820 Chr.12: 6728289-6728289 on Build GRCh38
rs903359857 Chr.12: 6728183-6728183 on Build GRCh38
rs914796608 Chr.12: 6728154-6728154 on Build GRCh38
rs772978808 Chr.12: 6728208-6728208 on Build GRCh38
rs376745830 Chr.12: 6728204-6728204 on Build GRCh38
rs749811660 Chr.12: 6728218-6728218 on Build GRCh38
rs756944860 Chr.12: 6728212-6728212 on Build GRCh38
rs372627223 Chr.12: 6728184-6728184 on Build GRCh38
rs754479824 Chr.12: 6728324-6728324 on Build GRCh38
rs758306221 Chr.12: 6728306-6728306 on Build GRCh38
rs931975184 Chr.12: 6728115-6728115 on Build GRCh38
rs749977841 Chr.12: 6728202-6728202 on Build GRCh38
rs756993027 Chr.12: 6728190-6728190 on Build GRCh38
rs749418560 Chr.12: 6728316-6728316 on Build GRCh38
rs1051710156 Chr.12: 6728293-6728293 on Build GRCh38
rs888767407 Chr.12: 6728311-6728311 on Build GRCh38
rs1041866627 Chr.12: 6728165-6728165 on Build GRCh38
rs1006097754 Chr.12: 6728254-6728254 on Build GRCh38
rs373756883 Chr.12: 6728211-6728211 on Build GRCh38
rs764220895 Chr.12: 6728274-6728274 on Build GRCh38
rs114836259 Chr.12: 6728233-6728233 on Build GRCh38
rs764902554 Chr.12: 6728280-6728280 on Build GRCh38
rs141811786 Chr.12: 6728223-6728223 on Build GRCh38
rs761815242 Chr.12: 6728277-6728277 on Build GRCh38
rs779847259 Chr.12: 6728314-6728314 on Build GRCh38
rs759703229 Chr.12: 6728243-6728243 on Build GRCh38
rs772262508 Chr.12: 6728245-6728245 on Build GRCh38
rs750256819 Chr.12: 6728290-6728290 on Build GRCh38
rs369439337 Chr.12: 6728210-6728210 on Build GRCh38
rs775733098 Chr.12: 6728262-6728262 on Build GRCh38

View Full Product Details