Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35555623-35555862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4A
Entrez Gene ID
495
Gene Name
ATPase H+/K+ transporting alpha subunit
Gene Aliases
ATP6A
UniGene
Hs.36992
Gene Chromosome Location
Chr.19: 35550193-35563658 on Build GRCh38
Transcripts
NM_000704.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607918 c.1928G>A p.G643D Chr.19: 35555754-35555754 on Build GRCh38
1245748 c.1987G>A p.V663I Chr.19: 35555695-35555695 on Build GRCh38
3672856 c.1884G>T p.T628T Chr.19: 35555798-35555798 on Build GRCh38
4664408 c.1876A>C p.M626L Chr.19: 35555806-35555806 on Build GRCh38
4565341 c.1992_1993CC>TT p.Q665* Chr.19: 35555689-35555690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769485930 Chr.19: 35555663-35555664 on Build GRCh38
rs375470351 Chr.19: 35555817-35555817 on Build GRCh38
rs372060721 Chr.19: 35555780-35555780 on Build GRCh38
rs80198561 Chr.19: 35555665-35555665 on Build GRCh38
rs753426264 Chr.19: 35555735-35555735 on Build GRCh38
rs267605431 Chr.19: 35555692-35555692 on Build GRCh38
rs757758386 Chr.19: 35555704-35555704 on Build GRCh38
rs61736770 Chr.19: 35555696-35555696 on Build GRCh38
rs768008534 Chr.19: 35555829-35555829 on Build GRCh38
rs778879380 Chr.19: 35555773-35555773 on Build GRCh38
rs1007249420 Chr.19: 35555778-35555778 on Build GRCh38
rs755734977 Chr.19: 35555726-35555726 on Build GRCh38
rs775065728 Chr.19: 35555824-35555824 on Build GRCh38
rs767990060 Chr.19: 35555710-35555710 on Build GRCh38
rs752094325 Chr.19: 35555703-35555703 on Build GRCh38
rs145767701 Chr.19: 35555679-35555679 on Build GRCh38
rs775546496 Chr.19: 35555814-35555814 on Build GRCh38
rs770021144 Chr.19: 35555795-35555795 on Build GRCh38
rs748068530 Chr.19: 35555777-35555777 on Build GRCh38
rs1023068872 Chr.19: 35555798-35555798 on Build GRCh38
rs754606914 Chr.19: 35555825-35555825 on Build GRCh38
rs368721895 Chr.19: 35555728-35555728 on Build GRCh38
rs967405578 Chr.19: 35555657-35555657 on Build GRCh38
rs888887677 Chr.19: 35555770-35555770 on Build GRCh38
rs989838077 Chr.19: 35555661-35555661 on Build GRCh38
rs1028106478 Chr.19: 35555755-35555755 on Build GRCh38
rs138654635 Chr.19: 35555680-35555680 on Build GRCh38
rs763145432 Chr.19: 35555815-35555815 on Build GRCh38
rs898108443 Chr.19: 35555709-35555709 on Build GRCh38
rs768595874 Chr.19: 35555653-35555653 on Build GRCh38
rs763403043 Chr.19: 35555687-35555687 on Build GRCh38
rs774239291 Chr.19: 35555654-35555654 on Build GRCh38
rs201830604 Chr.19: 35555717-35555717 on Build GRCh38
rs370554663 Chr.19: 35555670-35555670 on Build GRCh38
rs779455265 Chr.19: 35555734-35555734 on Build GRCh38
rs746098310 Chr.19: 35555786-35555786 on Build GRCh38
rs777563260 Chr.19: 35555783-35555783 on Build GRCh38
rs775544857 Chr.19: 35555664-35555664 on Build GRCh38
rs150371575 Chr.19: 35555745-35555745 on Build GRCh38

View Full Product Details