Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13757307-13757578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC130
Entrez Gene ID
81576
Gene Name
coiled-coil domain containing 130
Gene Aliases
-
UniGene
Hs.24998
Gene Chromosome Location
Chr.19: 13731760-13763298 on Build GRCh38
Transcripts
NM_001320561.1 NM_001320564.1 NM_001320565.1 NM_001320566.1 NM_001320567.1 NM_001320568.1 NM_001320569.1 NM_030818.3 XM_005260086.4 XM_011528326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
991726 c.144C>T p.F48F Chr.19: 13757421-13757421 on Build GRCh38
991727 c.168C>T p.C56C Chr.19: 13757445-13757445 on Build GRCh38
3529216 c.184C>T p.H62Y Chr.19: 13757461-13757461 on Build GRCh38
2818000 c.190G>A p.G64S Chr.19: 13757467-13757467 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050353744 Chr.19: 13757545-13757545 on Build GRCh38
rs540909958 Chr.19: 13757448-13757448 on Build GRCh38
rs532671223 Chr.19: 13757400-13757400 on Build GRCh38
rs536269729 Chr.19: 13757365-13757365 on Build GRCh38
rs202065091 Chr.19: 13757467-13757467 on Build GRCh38
rs372094950 Chr.19: 13757489-13757489 on Build GRCh38
rs548139377 Chr.19: 13757350-13757350 on Build GRCh38
rs754123656 Chr.19: 13757443-13757443 on Build GRCh38
rs935934132 Chr.19: 13757543-13757543 on Build GRCh38
rs1042020033 Chr.19: 13757340-13757340 on Build GRCh38
rs140514061 Chr.19: 13757464-13757464 on Build GRCh38
rs374992243 Chr.19: 13757446-13757446 on Build GRCh38
rs755753555 Chr.19: 13757473-13757473 on Build GRCh38
rs779735921 Chr.19: 13757390-13757390 on Build GRCh38
rs760833880 Chr.19: 13757421-13757421 on Build GRCh38
rs1056931235 Chr.19: 13757329-13757329 on Build GRCh38
rs199653797 Chr.19: 13757393-13757393 on Build GRCh38
rs755882745 Chr.19: 13757480-13757481 on Build GRCh38
rs779862619 Chr.19: 13757485-13757485 on Build GRCh38
rs945942912 Chr.19: 13757339-13757339 on Build GRCh38
rs371118275 Chr.19: 13757420-13757420 on Build GRCh38
rs756689526 Chr.19: 13757466-13757466 on Build GRCh38
rs772217178 Chr.19: 13757515-13757515 on Build GRCh38
rs750171776 Chr.19: 13757369-13757370 on Build GRCh38
rs768550633 Chr.19: 13757482-13757482 on Build GRCh38
rs373911398 Chr.19: 13757341-13757341 on Build GRCh38
rs750042354 Chr.19: 13757471-13757471 on Build GRCh38
rs904426454 Chr.19: 13757541-13757541 on Build GRCh38
rs113249008 Chr.19: 13757537-13757537 on Build GRCh38
rs779794634 Chr.19: 13757474-13757474 on Build GRCh38
rs575036358 Chr.19: 13757369-13757369 on Build GRCh38
rs963015001 Chr.19: 13757548-13757548 on Build GRCh38
rs937036387 Chr.19: 13757492-13757492 on Build GRCh38
rs1027887956 Chr.19: 13757461-13757461 on Build GRCh38
rs992986553 Chr.19: 13757493-13757493 on Build GRCh38
rs75450630 Chr.19: 13757357-13757357 on Build GRCh38
rs576654636 Chr.19: 13757433-13757433 on Build GRCh38
rs759832662 Chr.19: 13757500-13757500 on Build GRCh38
rs895642738 Chr.19: 13757338-13757338 on Build GRCh38
rs773461190 Chr.19: 13757518-13757518 on Build GRCh38
rs945859876 Chr.19: 13757520-13757520 on Build GRCh38
rs776660538 Chr.19: 13757459-13757459 on Build GRCh38
rs749013073 Chr.19: 13757479-13757479 on Build GRCh38
rs1010596660 Chr.19: 13757402-13757405 on Build GRCh38
rs199876154 Chr.19: 13757415-13757415 on Build GRCh38
rs374186119 Chr.19: 13757354-13757354 on Build GRCh38
rs537187967 Chr.19: 13757411-13757411 on Build GRCh38
rs148234365 Chr.19: 13757546-13757546 on Build GRCh38
rs1000134359 Chr.19: 13757351-13757351 on Build GRCh38
rs889112417 Chr.19: 13757551-13757551 on Build GRCh38

View Full Product Details