Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49417533-49417788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMT
Entrez Gene ID
275
Gene Name
aminomethyltransferase
Gene Aliases
GCE, GCST, GCVT, NKH
UniGene
Hs.102
Gene Chromosome Location
Chr.3: 49416778-49422678 on Build GRCh38
Transcripts
NM_000481.3 NM_001164710.1 NM_001164711.1 NM_001164712.1 NR_028435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2849906 c.1082C>T p.A361V Chr.3: 49417670-49417670 on Build GRCh38
2849904 c.1155G>C p.Q385H Chr.3: 49417597-49417597 on Build GRCh38
6228150 c.1123A>G p.M375V Chr.3: 49417629-49417629 on Build GRCh38
2849903 c.1177C>T p.P393S Chr.3: 49417575-49417575 on Build GRCh38
1046149 c.1112G>A p.R371H Chr.3: 49417640-49417640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775324601 Chr.3: 49417611-49417611 on Build GRCh38
rs377179216 Chr.3: 49417556-49417556 on Build GRCh38
rs886058683 Chr.3: 49417562-49417562 on Build GRCh38
rs747901545 Chr.3: 49417699-49417699 on Build GRCh38
rs370432325 Chr.3: 49417750-49417750 on Build GRCh38
rs532787574 Chr.3: 49417714-49417714 on Build GRCh38
rs768293167 Chr.3: 49417694-49417694 on Build GRCh38
rs750350027 Chr.3: 49417689-49417689 on Build GRCh38
rs373269572 Chr.3: 49417754-49417754 on Build GRCh38
rs780708903 Chr.3: 49417695-49417695 on Build GRCh38
rs762157837 Chr.3: 49417752-49417752 on Build GRCh38
rs756452579 Chr.3: 49417555-49417555 on Build GRCh38
rs779034974 Chr.3: 49417683-49417683 on Build GRCh38
rs760960965 Chr.3: 49417746-49417746 on Build GRCh38
rs777754811 Chr.3: 49417720-49417720 on Build GRCh38
rs147006017 Chr.3: 49417640-49417640 on Build GRCh38
rs751298230 Chr.3: 49417765-49417765 on Build GRCh38
rs769323684 Chr.3: 49417695-49417695 on Build GRCh38
rs760912937 Chr.3: 49417653-49417653 on Build GRCh38
rs748603289 Chr.3: 49417689-49417690 on Build GRCh38
rs779781655 Chr.3: 49417691-49417691 on Build GRCh38
rs772953192 Chr.3: 49417699-49417700 on Build GRCh38
rs757293245 Chr.3: 49417670-49417670 on Build GRCh38
rs764310045 Chr.3: 49417748-49417748 on Build GRCh38
rs757170367 Chr.3: 49417595-49417595 on Build GRCh38
rs776872417 Chr.3: 49417689-49417689 on Build GRCh38
rs141246107 Chr.3: 49417607-49417607 on Build GRCh38
rs775949489 Chr.3: 49417745-49417745 on Build GRCh38
rs201400089 Chr.3: 49417655-49417655 on Build GRCh38
rs777880082 Chr.3: 49417586-49417586 on Build GRCh38
rs200180779 Chr.3: 49417565-49417565 on Build GRCh38
rs753150990 Chr.3: 49417651-49417651 on Build GRCh38
rs377227163 Chr.3: 49417690-49417690 on Build GRCh38
rs760086617 Chr.3: 49417737-49417737 on Build GRCh38
rs768222025 Chr.3: 49417615-49417615 on Build GRCh38
rs768940499 Chr.3: 49417676-49417678 on Build GRCh38
rs201189946 Chr.3: 49417616-49417616 on Build GRCh38
rs373140736 Chr.3: 49417650-49417650 on Build GRCh38
rs771853703 Chr.3: 49417610-49417610 on Build GRCh38
rs746645414 Chr.3: 49417612-49417612 on Build GRCh38
rs754720687 Chr.3: 49417769-49417769 on Build GRCh38
rs561380533 Chr.3: 49417601-49417601 on Build GRCh38
rs758281017 Chr.3: 49417755-49417755 on Build GRCh38
rs972972652 Chr.3: 49417597-49417597 on Build GRCh38
rs571827952 Chr.3: 49417693-49417693 on Build GRCh38
rs772896226 Chr.3: 49417732-49417732 on Build GRCh38
rs753899057 Chr.3: 49417669-49417669 on Build GRCh38
rs1009661817 Chr.3: 49417729-49417729 on Build GRCh38
rs767969842 Chr.3: 49417648-49417648 on Build GRCh38
rs772738913 Chr.3: 49417740-49417740 on Build GRCh38
rs764246902 Chr.3: 49417660-49417660 on Build GRCh38
rs761239253 Chr.3: 49417627-49417627 on Build GRCh38
rs747544827 Chr.3: 49417597-49417599 on Build GRCh38
rs760178260 Chr.3: 49417646-49417646 on Build GRCh38
rs749359730 Chr.3: 49417590-49417590 on Build GRCh38
rs201548428 Chr.3: 49417553-49417553 on Build GRCh38
rs779977631 Chr.3: 49417761-49417761 on Build GRCh38

View Full Product Details