Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10306439-10306674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM
Entrez Gene ID
133
Gene Name
adrenomedullin
Gene Aliases
AM, PAMP
UniGene
Hs.441047
Gene Chromosome Location
Chr.11: 10304980-10307402 on Build GRCh38
Transcripts
NM_001124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
922375 c.518C>A p.A173D Chr.11: 10306601-10306601 on Build GRCh38
1350456 c.438C>T p.Y146Y Chr.11: 10306521-10306521 on Build GRCh38
7218854 c.531G>T p.P177P Chr.11: 10306614-10306614 on Build GRCh38
6306738 c.378G>T p.Q126H Chr.11: 10306461-10306461 on Build GRCh38
1350458 c.524delC p.P177fs*>9 Chr.11: 10306607-10306607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750283609 Chr.11: 10306559-10306559 on Build GRCh38
rs765070563 Chr.11: 10306555-10306555 on Build GRCh38
rs775154162 Chr.11: 10306540-10306540 on Build GRCh38
rs770133350 Chr.11: 10306515-10306515 on Build GRCh38
rs750807199 Chr.11: 10306650-10306650 on Build GRCh38
rs755194034 Chr.11: 10306573-10306573 on Build GRCh38
rs781376753 Chr.11: 10306513-10306513 on Build GRCh38
rs140844268 Chr.11: 10306468-10306468 on Build GRCh38
rs746665872 Chr.11: 10306636-10306636 on Build GRCh38
rs199526740 Chr.11: 10306592-10306592 on Build GRCh38
rs988335968 Chr.11: 10306598-10306598 on Build GRCh38
rs112526486 Chr.11: 10306461-10306461 on Build GRCh38
rs529300345 Chr.11: 10306486-10306486 on Build GRCh38
rs371681360 Chr.11: 10306499-10306499 on Build GRCh38
rs374900367 Chr.11: 10306581-10306581 on Build GRCh38
rs778109412 Chr.11: 10306611-10306611 on Build GRCh38
rs373720232 Chr.11: 10306612-10306612 on Build GRCh38
rs748703275 Chr.11: 10306514-10306514 on Build GRCh38
rs773405081 Chr.11: 10306606-10306607 on Build GRCh38
rs773695276 Chr.11: 10306522-10306522 on Build GRCh38
rs866419823 Chr.11: 10306517-10306517 on Build GRCh38
rs150162311 Chr.11: 10306479-10306479 on Build GRCh38
rs373588240 Chr.11: 10306610-10306610 on Build GRCh38
rs763603296 Chr.11: 10306546-10306546 on Build GRCh38
rs761874343 Chr.11: 10306552-10306552 on Build GRCh38
rs771983072 Chr.11: 10306478-10306485 on Build GRCh38
rs774639478 Chr.11: 10306623-10306623 on Build GRCh38
rs926266115 Chr.11: 10306631-10306631 on Build GRCh38
rs751857285 Chr.11: 10306562-10306562 on Build GRCh38
rs778770096 Chr.11: 10306480-10306480 on Build GRCh38
rs747129027 Chr.11: 10306501-10306501 on Build GRCh38
rs771749755 Chr.11: 10306537-10306537 on Build GRCh38
rs763554599 Chr.11: 10306466-10306466 on Build GRCh38
rs753216941 Chr.11: 10306548-10306548 on Build GRCh38
rs768891270 Chr.11: 10306512-10306512 on Build GRCh38
rs763215841 Chr.11: 10306524-10306524 on Build GRCh38
rs748173146 Chr.11: 10306609-10306609 on Build GRCh38
rs371598515 Chr.11: 10306560-10306560 on Build GRCh38
rs758702560 Chr.11: 10306491-10306491 on Build GRCh38
rs760271378 Chr.11: 10306542-10306542 on Build GRCh38
rs776259273 Chr.11: 10306652-10306652 on Build GRCh38
rs771306099 Chr.11: 10306616-10306616 on Build GRCh38
rs781336657 Chr.11: 10306603-10306603 on Build GRCh38

View Full Product Details