Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120254782-120255013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM255A
Entrez Gene ID
55026
Gene Name
transmembrane protein 255A
Gene Aliases
FAM70A
UniGene
Hs.437563
Gene Chromosome Location
Chr.X: 120250984-120311556 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

Gene Symbol
ZBTB33
Entrez Gene ID
10009
Gene Name
zinc finger and BTB domain containing 33
Gene Aliases
ZNF-kaiso, ZNF348
UniGene
Hs.143604
Gene Chromosome Location
Chr.X: 120250752-120258396 on Build GRCh38
Transcripts
XM_017029619.1 NM_001184742.1 NM_006777.3

SNP Detail

SNP ID Chromosome Location
rs782372203 Chr.X: 120254834-120254834 on Build GRCh38
rs782272539 Chr.X: 120254813-120254813 on Build GRCh38
rs782757193 Chr.X: 120254888-120254888 on Build GRCh38
rs782646752 Chr.X: 120254917-120254917 on Build GRCh38
rs782439868 Chr.X: 120254979-120254979 on Build GRCh38
rs782120191 Chr.X: 120254874-120254874 on Build GRCh38
rs150678838 Chr.X: 120254984-120254984 on Build GRCh38
rs201556300 Chr.X: 120254841-120254841 on Build GRCh38
rs782343981 Chr.X: 120254942-120254942 on Build GRCh38
rs1024614561 Chr.X: 120254959-120254959 on Build GRCh38
rs781900955 Chr.X: 120254900-120254900 on Build GRCh38
rs781874187 Chr.X: 120254861-120254861 on Build GRCh38
rs782464432 Chr.X: 120254896-120254896 on Build GRCh38
rs371846102 Chr.X: 120254822-120254822 on Build GRCh38
rs782086829 Chr.X: 120254823-120254823 on Build GRCh38
rs782029088 Chr.X: 120254812-120254812 on Build GRCh38
rs781817851 Chr.X: 120254893-120254893 on Build GRCh38
rs782184428 Chr.X: 120254968-120254968 on Build GRCh38
rs782385890 Chr.X: 120254817-120254817 on Build GRCh38
rs370321662 Chr.X: 120254897-120254897 on Build GRCh38
rs782241523 Chr.X: 120254922-120254922 on Build GRCh38
rs782528537 Chr.X: 120254902-120254902 on Build GRCh38
rs782168421 Chr.X: 120254839-120254839 on Build GRCh38
rs990511360 Chr.X: 120254923-120254923 on Build GRCh38
rs782134592 Chr.X: 120254988-120254988 on Build GRCh38
rs143194661 Chr.X: 120254825-120254825 on Build GRCh38
rs782598959 Chr.X: 120254957-120254957 on Build GRCh38

View Full Product Details