Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50463725-50463998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF2
Entrez Gene ID
80221
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
ACSMW, AVYV493
UniGene
Hs.288959
Gene Chromosome Location
Chr.17: 50426158-50474845 on Build GRCh38
Transcripts
NM_001288968.1 NM_001288969.1 NM_001288970.1 NM_001288971.1 NM_001288972.1 NM_025149.5 XM_006722110.3 XM_011525294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726681 c.1052C>T p.T351I Chr.17: 50463823-50463823 on Build GRCh38
5620197 c.1060T>C p.Y354H Chr.17: 50463831-50463831 on Build GRCh38
1479722 c.1125G>A p.S375S Chr.17: 50463896-50463896 on Build GRCh38
6592535 c.1079T>C p.F360S Chr.17: 50463850-50463850 on Build GRCh38
3889886 c.1080C>T p.F360F Chr.17: 50463851-50463851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753090663 Chr.17: 50463842-50463842 on Build GRCh38
rs762883572 Chr.17: 50463814-50463814 on Build GRCh38
rs760961595 Chr.17: 50463959-50463959 on Build GRCh38
rs184135677 Chr.17: 50463951-50463951 on Build GRCh38
rs754285481 Chr.17: 50463961-50463961 on Build GRCh38
rs755173170 Chr.17: 50463770-50463770 on Build GRCh38
rs773202679 Chr.17: 50463813-50463813 on Build GRCh38
rs924198512 Chr.17: 50463750-50463750 on Build GRCh38
rs375128217 Chr.17: 50463898-50463898 on Build GRCh38
rs769759679 Chr.17: 50463812-50463812 on Build GRCh38
rs34471056 Chr.17: 50463828-50463829 on Build GRCh38
rs536922711 Chr.17: 50463973-50463973 on Build GRCh38
rs747971781 Chr.17: 50463912-50463912 on Build GRCh38
rs878898810 Chr.17: 50463755-50463755 on Build GRCh38
rs756616605 Chr.17: 50463780-50463780 on Build GRCh38
rs200992227 Chr.17: 50463821-50463821 on Build GRCh38
rs771638590 Chr.17: 50463789-50463789 on Build GRCh38
rs563258697 Chr.17: 50463765-50463765 on Build GRCh38
rs746622866 Chr.17: 50463798-50463798 on Build GRCh38
rs992587609 Chr.17: 50463902-50463902 on Build GRCh38
rs778283986 Chr.17: 50463847-50463847 on Build GRCh38
rs768793712 Chr.17: 50463954-50463955 on Build GRCh38
rs780390434 Chr.17: 50463778-50463779 on Build GRCh38
rs375884059 Chr.17: 50463896-50463896 on Build GRCh38
rs770890963 Chr.17: 50463942-50463942 on Build GRCh38
rs371395948 Chr.17: 50463799-50463799 on Build GRCh38
rs200056965 Chr.17: 50463857-50463857 on Build GRCh38
rs890158349 Chr.17: 50463867-50463867 on Build GRCh38
rs778480260 Chr.17: 50463782-50463782 on Build GRCh38
rs936413365 Chr.17: 50463759-50463759 on Build GRCh38
rs779669695 Chr.17: 50463860-50463860 on Build GRCh38
rs917878782 Chr.17: 50463917-50463917 on Build GRCh38
rs753037399 Chr.17: 50463776-50463776 on Build GRCh38
rs994746636 Chr.17: 50463914-50463914 on Build GRCh38
rs368079198 Chr.17: 50463804-50463804 on Build GRCh38
rs552297060 Chr.17: 50463803-50463803 on Build GRCh38
rs962866951 Chr.17: 50463832-50463832 on Build GRCh38
rs768330040 Chr.17: 50463895-50463895 on Build GRCh38
rs767675694 Chr.17: 50463833-50463833 on Build GRCh38
rs757860734 Chr.17: 50463852-50463852 on Build GRCh38
rs752793765 Chr.17: 50463954-50463954 on Build GRCh38
rs781575468 Chr.17: 50463775-50463775 on Build GRCh38
rs745364524 Chr.17: 50463785-50463785 on Build GRCh38
rs144013435 Chr.17: 50463875-50463875 on Build GRCh38
rs752743919 Chr.17: 50463909-50463909 on Build GRCh38
rs201660426 Chr.17: 50463926-50463926 on Build GRCh38
rs762832072 Chr.17: 50463930-50463930 on Build GRCh38
rs139044093 Chr.17: 50463845-50463845 on Build GRCh38
rs751666003 Chr.17: 50463819-50463819 on Build GRCh38
rs779720563 Chr.17: 50463790-50463790 on Build GRCh38
rs754334661 Chr.17: 50463851-50463851 on Build GRCh38
rs759586377 Chr.17: 50463943-50463943 on Build GRCh38
rs759767064 Chr.17: 50463826-50463826 on Build GRCh38
rs746991801 Chr.17: 50463805-50463805 on Build GRCh38

View Full Product Details