Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10451348-10451621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APITD1-CORT
Entrez Gene ID
100526739
Gene Name
APITD1-CORT readthrough
Gene Aliases
-
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10452003 on Build GRCh38
Transcripts
NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1 NM_001302.4

Gene Symbol
CORT
Entrez Gene ID
1325
Gene Name
cortistatin
Gene Aliases
CST-14, CST-17, CST-29
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10449719-10452003 on Build GRCh38
Transcripts
NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1 NM_001302.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6120214 c.352C>G p.Q118E Chr.1: 10451452-10451452 on Build GRCh38
6533566 c.367C>G p.P123A Chr.1: 10451467-10451467 on Build GRCh38
6533567 c.340C>G p.P114A Chr.1: 10451467-10451467 on Build GRCh38
6602181 c.342C>T p.P114P Chr.1: 10451469-10451469 on Build GRCh38
5673895 c.404G>A p.R135Q Chr.1: 10451504-10451504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754668122 Chr.1: 10451544-10451544 on Build GRCh38
rs748969228 Chr.1: 10451518-10451518 on Build GRCh38
rs199777356 Chr.1: 10451504-10451504 on Build GRCh38
rs764155114 Chr.1: 10451534-10451534 on Build GRCh38
rs375132033 Chr.1: 10451463-10451463 on Build GRCh38
rs766189277 Chr.1: 10451530-10451530 on Build GRCh38
rs751627080 Chr.1: 10451503-10451503 on Build GRCh38
rs373837353 Chr.1: 10451390-10451390 on Build GRCh38
rs759210368 Chr.1: 10451579-10451579 on Build GRCh38
rs547659978 Chr.1: 10451404-10451404 on Build GRCh38
rs779616729 Chr.1: 10451514-10451514 on Build GRCh38
rs993756376 Chr.1: 10451591-10451591 on Build GRCh38
rs781697531 Chr.1: 10451428-10451428 on Build GRCh38
rs666103 Chr.1: 10451532-10451532 on Build GRCh38
rs776489624 Chr.1: 10451578-10451578 on Build GRCh38
rs757361401 Chr.1: 10451537-10451537 on Build GRCh38
rs962257726 Chr.1: 10451541-10451541 on Build GRCh38
rs779570883 Chr.1: 10451518-10451518 on Build GRCh38
rs375844912 Chr.1: 10451470-10451470 on Build GRCh38
rs916016644 Chr.1: 10451381-10451381 on Build GRCh38
rs768186124 Chr.1: 10451538-10451538 on Build GRCh38
rs774690087 Chr.1: 10451598-10451598 on Build GRCh38
rs760500441 Chr.1: 10451525-10451525 on Build GRCh38
rs878988813 Chr.1: 10451531-10451532 on Build GRCh38
rs935263406 Chr.1: 10451439-10451439 on Build GRCh38
rs773086446 Chr.1: 10451523-10451523 on Build GRCh38
rs41376048 Chr.1: 10451531-10451531 on Build GRCh38
rs756481798 Chr.1: 10451512-10451512 on Build GRCh38
rs747513324 Chr.1: 10451410-10451410 on Build GRCh38
rs550348685 Chr.1: 10451533-10451533 on Build GRCh38
rs747591951 Chr.1: 10451521-10451521 on Build GRCh38
rs769598738 Chr.1: 10451443-10451443 on Build GRCh38
rs544551198 Chr.1: 10451489-10451489 on Build GRCh38
rs145148065 Chr.1: 10451539-10451539 on Build GRCh38
rs971817390 Chr.1: 10451536-10451536 on Build GRCh38
rs775156999 Chr.1: 10451460-10451460 on Build GRCh38
rs75501939 Chr.1: 10451524-10451524 on Build GRCh38
rs778182842 Chr.1: 10451408-10451408 on Build GRCh38
rs771479464 Chr.1: 10451424-10451424 on Build GRCh38
rs906613670 Chr.1: 10451476-10451476 on Build GRCh38
rs748612792 Chr.1: 10451570-10451570 on Build GRCh38
rs774224705 Chr.1: 10451465-10451465 on Build GRCh38
rs767385934 Chr.1: 10451474-10451474 on Build GRCh38
rs746165625 Chr.1: 10451430-10451430 on Build GRCh38
rs773865457 Chr.1: 10451520-10451520 on Build GRCh38
rs764397874 Chr.1: 10451592-10451592 on Build GRCh38
rs772029734 Chr.1: 10451522-10451522 on Build GRCh38
rs894406784 Chr.1: 10451432-10451432 on Build GRCh38
rs906436872 Chr.1: 10451427-10451427 on Build GRCh38
rs367676737 Chr.1: 10451493-10451493 on Build GRCh38
rs746254364 Chr.1: 10451511-10451511 on Build GRCh38
rs628462 Chr.1: 10451487-10451487 on Build GRCh38
rs770711751 Chr.1: 10451564-10451564 on Build GRCh38
rs768113640 Chr.1: 10451519-10451519 on Build GRCh38
rs1027949824 Chr.1: 10451594-10451594 on Build GRCh38
rs201163116 Chr.1: 10451477-10451477 on Build GRCh38
rs372806878 Chr.1: 10451376-10451376 on Build GRCh38
rs200835553 Chr.1: 10451488-10451488 on Build GRCh38
rs893474399 Chr.1: 10451478-10451478 on Build GRCh38
rs752753240 Chr.1: 10451399-10451399 on Build GRCh38
rs991588592 Chr.1: 10451380-10451380 on Build GRCh38
rs201158337 Chr.1: 10451464-10451464 on Build GRCh38
rs773198534 Chr.1: 10451552-10451552 on Build GRCh38
rs755207510 Chr.1: 10451382-10451382 on Build GRCh38
rs746947877 Chr.1: 10451563-10451563 on Build GRCh38
rs765425169 Chr.1: 10451481-10451481 on Build GRCh38
rs772618348 Chr.1: 10451580-10451582 on Build GRCh38
rs749561966 Chr.1: 10451542-10451542 on Build GRCh38
rs780453865 Chr.1: 10451540-10451540 on Build GRCh38
rs751718989 Chr.1: 10451535-10451535 on Build GRCh38
rs755613959 Chr.1: 10451517-10451518 on Build GRCh38
rs571865438 Chr.1: 10451549-10451549 on Build GRCh38
rs747682104 Chr.1: 10451547-10451547 on Build GRCh38

View Full Product Details