Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 86262699-86262960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
UniGene
Hs.31795
Gene Chromosome Location
Chr.15: 86079635-87031476 on Build GRCh38
Transcripts
XM_011521226.2 XM_011521227.2 XM_011521228.2 XM_011521230.2 XM_017021918.1 XM_017021919.1 XM_017021920.1 XM_017021921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6532143 c.56T>C p.L19P Chr.15: 86262809-86262809 on Build GRCh38
4416259 c.852C>T p.D284D Chr.15: 86262798-86262798 on Build GRCh38
6532142 c.863T>C p.L288P Chr.15: 86262809-86262809 on Build GRCh38
1493561 c.846A>G p.E282E Chr.15: 86262792-86262792 on Build GRCh38
3387124 c.79C>T p.R27* Chr.15: 86262832-86262832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894736461 Chr.15: 86262924-86262924 on Build GRCh38
rs763558239 Chr.15: 86262805-86262805 on Build GRCh38
rs772567921 Chr.15: 86262934-86262934 on Build GRCh38
rs746918783 Chr.15: 86262895-86262895 on Build GRCh38
rs1013480040 Chr.15: 86262735-86262735 on Build GRCh38
rs944575453 Chr.15: 86262852-86262852 on Build GRCh38
rs143754366 Chr.15: 86262864-86262864 on Build GRCh38
rs769765314 Chr.15: 86262912-86262912 on Build GRCh38
rs757272017 Chr.15: 86262899-86262899 on Build GRCh38
rs371826143 Chr.15: 86262861-86262861 on Build GRCh38
rs767815188 Chr.15: 86262817-86262817 on Build GRCh38
rs904907838 Chr.15: 86262777-86262777 on Build GRCh38
rs1566088 Chr.15: 86262798-86262798 on Build GRCh38
rs761802320 Chr.15: 86262858-86262858 on Build GRCh38
rs200285527 Chr.15: 86262832-86262832 on Build GRCh38
rs772825413 Chr.15: 86262911-86262911 on Build GRCh38
rs773750005 Chr.15: 86262928-86262928 on Build GRCh38
rs557218142 Chr.15: 86262807-86262807 on Build GRCh38
rs971744826 Chr.15: 86262823-86262823 on Build GRCh38
rs746961089 Chr.15: 86262774-86262774 on Build GRCh38
rs924728917 Chr.15: 86262874-86262874 on Build GRCh38
rs777745123 Chr.15: 86262771-86262771 on Build GRCh38
rs770812651 Chr.15: 86262790-86262790 on Build GRCh38
rs575477962 Chr.15: 86262822-86262822 on Build GRCh38
rs542945408 Chr.15: 86262927-86262927 on Build GRCh38
rs762255401 Chr.15: 86262808-86262808 on Build GRCh38
rs1013127063 Chr.15: 86262933-86262933 on Build GRCh38
rs989397146 Chr.15: 86262734-86262734 on Build GRCh38
rs994834979 Chr.15: 86262906-86262906 on Build GRCh38
rs561280315 Chr.15: 86262937-86262937 on Build GRCh38
rs893362547 Chr.15: 86262921-86262921 on Build GRCh38
rs755055710 Chr.15: 86262855-86262855 on Build GRCh38
rs775181651 Chr.15: 86262917-86262917 on Build GRCh38
rs986334707 Chr.15: 86262839-86262839 on Build GRCh38
rs776588646 Chr.15: 86262792-86262792 on Build GRCh38
rs1050688457 Chr.15: 86262821-86262821 on Build GRCh38
rs35771845 Chr.15: 86262830-86262830 on Build GRCh38
rs551065804 Chr.15: 86262932-86262932 on Build GRCh38
rs762507639 Chr.15: 86262804-86262804 on Build GRCh38
rs781066217 Chr.15: 86262907-86262907 on Build GRCh38
rs745458688 Chr.15: 86262793-86262793 on Build GRCh38
rs951955069 Chr.15: 86262833-86262833 on Build GRCh38
rs878918020 Chr.15: 86262913-86262913 on Build GRCh38
rs754056355 Chr.15: 86262844-86262844 on Build GRCh38
rs748929540 Chr.15: 86262926-86262926 on Build GRCh38
rs866857305 Chr.15: 86262867-86262867 on Build GRCh38
rs200028178 Chr.15: 86262851-86262851 on Build GRCh38
rs140123227 Chr.15: 86262770-86262770 on Build GRCh38

View Full Product Details