Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26582710-26582969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB4
Entrez Gene ID
130106
Gene Name
calcium and integrin binding family member 4
Gene Aliases
KIP4
UniGene
Hs.591579
Gene Chromosome Location
Chr.2: 26581202-26641798 on Build GRCh38
Transcripts
NM_001029881.2 XM_011532514.2 XM_011532517.2 XM_017003329.1 XM_017003330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5465523 c.457C>T p.L153L Chr.2: 26582895-26582895 on Build GRCh38
4962120 c.485C>G p.S162* Chr.2: 26582867-26582867 on Build GRCh38
1019428 c.525G>T p.M175I Chr.2: 26582827-26582827 on Build GRCh38
6835515 c.487G>A p.E163K Chr.2: 26582865-26582865 on Build GRCh38
6880580 c.527+2T>A p.? Chr.2: 26582823-26582823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750933335 Chr.2: 26582885-26582885 on Build GRCh38
rs373792460 Chr.2: 26582787-26582787 on Build GRCh38
rs768248734 Chr.2: 26582807-26582807 on Build GRCh38
rs568657123 Chr.2: 26582751-26582751 on Build GRCh38
rs747393783 Chr.2: 26582800-26582800 on Build GRCh38
rs7595964 Chr.2: 26582893-26582893 on Build GRCh38
rs778044733 Chr.2: 26582777-26582777 on Build GRCh38
rs759281760 Chr.2: 26582938-26582938 on Build GRCh38
rs997935114 Chr.2: 26582926-26582926 on Build GRCh38
rs760728601 Chr.2: 26582784-26582784 on Build GRCh38
rs749936526 Chr.2: 26582798-26582798 on Build GRCh38
rs372638135 Chr.2: 26582918-26582918 on Build GRCh38
rs994165847 Chr.2: 26582805-26582805 on Build GRCh38
rs202002208 Chr.2: 26582909-26582909 on Build GRCh38
rs370969445 Chr.2: 26582900-26582900 on Build GRCh38
rs775424318 Chr.2: 26582778-26582778 on Build GRCh38
rs142991987 Chr.2: 26582824-26582824 on Build GRCh38
rs141074863 Chr.2: 26582904-26582904 on Build GRCh38
rs368725686 Chr.2: 26582899-26582899 on Build GRCh38
rs780329569 Chr.2: 26582799-26582799 on Build GRCh38
rs144620649 Chr.2: 26582849-26582849 on Build GRCh38
rs780668047 Chr.2: 26582758-26582758 on Build GRCh38
rs753916098 Chr.2: 26582778-26582778 on Build GRCh38
rs2289703 Chr.2: 26582808-26582808 on Build GRCh38
rs201379980 Chr.2: 26582889-26582889 on Build GRCh38
rs767077695 Chr.2: 26582744-26582744 on Build GRCh38
rs776085882 Chr.2: 26582842-26582843 on Build GRCh38
rs928379931 Chr.2: 26582841-26582841 on Build GRCh38
rs374989653 Chr.2: 26582855-26582855 on Build GRCh38
rs768416269 Chr.2: 26582783-26582784 on Build GRCh38
rs888017056 Chr.2: 26582881-26582881 on Build GRCh38
rs754051621 Chr.2: 26582864-26582864 on Build GRCh38
rs138527374 Chr.2: 26582906-26582906 on Build GRCh38
rs767294998 Chr.2: 26582943-26582943 on Build GRCh38
rs766042141 Chr.2: 26582890-26582890 on Build GRCh38
rs150008757 Chr.2: 26582875-26582875 on Build GRCh38
rs991498411 Chr.2: 26582823-26582823 on Build GRCh38
rs774401469 Chr.2: 26582937-26582937 on Build GRCh38
rs771043666 Chr.2: 26582919-26582919 on Build GRCh38
rs747379720 Chr.2: 26582896-26582896 on Build GRCh38
rs147865735 Chr.2: 26582852-26582852 on Build GRCh38
rs764263564 Chr.2: 26582861-26582861 on Build GRCh38
rs773299975 Chr.2: 26582830-26582830 on Build GRCh38
rs773026292 Chr.2: 26582947-26582947 on Build GRCh38
rs914925499 Chr.2: 26582934-26582934 on Build GRCh38
rs373137228 Chr.2: 26582944-26582944 on Build GRCh38
rs755335625 Chr.2: 26582898-26582898 on Build GRCh38
rs902646467 Chr.2: 26582945-26582945 on Build GRCh38
rs540939424 Chr.2: 26582782-26582782 on Build GRCh38
rs181242068 Chr.2: 26582771-26582771 on Build GRCh38
rs1019665141 Chr.2: 26582768-26582768 on Build GRCh38
rs566343224 Chr.2: 26582842-26582842 on Build GRCh38

View Full Product Details