Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78573065-78573338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA3
Entrez Gene ID
306
Gene Name
annexin A3
Gene Aliases
ANX3
UniGene
Hs.480042
Gene Chromosome Location
Chr.4: 78551588-78610451 on Build GRCh38
Transcripts
NM_005139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601313 c.57C>A p.S19R Chr.4: 78573221-78573221 on Build GRCh38
225195 c.59C>T p.P20L Chr.4: 78573223-78573223 on Build GRCh38
6601314 c.26G>A p.R9Q Chr.4: 78573190-78573190 on Build GRCh38
4913575 c.70G>T p.A24S Chr.4: 78573234-78573234 on Build GRCh38
5815646 c.75A>T p.E25D Chr.4: 78573239-78573239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754956769 Chr.4: 78573190-78573190 on Build GRCh38
rs115874129 Chr.4: 78573094-78573094 on Build GRCh38
rs751792818 Chr.4: 78573306-78573306 on Build GRCh38
rs760325628 Chr.4: 78573211-78573211 on Build GRCh38
rs776925223 Chr.4: 78573264-78573264 on Build GRCh38
rs760071644 Chr.4: 78573268-78573268 on Build GRCh38
rs758611382 Chr.4: 78573117-78573117 on Build GRCh38
rs5951 Chr.4: 78573220-78573220 on Build GRCh38
rs950662837 Chr.4: 78573244-78573244 on Build GRCh38
rs745313749 Chr.4: 78573139-78573139 on Build GRCh38
rs376858661 Chr.4: 78573088-78573088 on Build GRCh38
rs774182367 Chr.4: 78573240-78573240 on Build GRCh38
rs545741268 Chr.4: 78573300-78573300 on Build GRCh38
rs778955878 Chr.4: 78573107-78573107 on Build GRCh38
rs745825275 Chr.4: 78573222-78573222 on Build GRCh38
rs370193019 Chr.4: 78573168-78573168 on Build GRCh38
rs776580900 Chr.4: 78573136-78573136 on Build GRCh38
rs560064715 Chr.4: 78573290-78573290 on Build GRCh38
rs757065040 Chr.4: 78573212-78573212 on Build GRCh38
rs369342841 Chr.4: 78573110-78573110 on Build GRCh38
rs766865529 Chr.4: 78573255-78573255 on Build GRCh38
rs746766118 Chr.4: 78573210-78573210 on Build GRCh38
rs756977230 Chr.4: 78573308-78573308 on Build GRCh38
rs201433481 Chr.4: 78573157-78573157 on Build GRCh38
rs994669293 Chr.4: 78573225-78573225 on Build GRCh38
rs768396844 Chr.4: 78573230-78573230 on Build GRCh38
rs761076043 Chr.4: 78573243-78573243 on Build GRCh38
rs759463168 Chr.4: 78573191-78573192 on Build GRCh38
rs757254118 Chr.4: 78573249-78573249 on Build GRCh38
rs1014311582 Chr.4: 78573213-78573213 on Build GRCh38
rs904759290 Chr.4: 78573163-78573163 on Build GRCh38
rs141337857 Chr.4: 78573189-78573189 on Build GRCh38
rs752773777 Chr.4: 78573193-78573193 on Build GRCh38
rs374243315 Chr.4: 78573197-78573197 on Build GRCh38
rs761166138 Chr.4: 78573185-78573185 on Build GRCh38
rs770823705 Chr.4: 78573119-78573119 on Build GRCh38
rs763898478 Chr.4: 78573167-78573167 on Build GRCh38
rs765353137 Chr.4: 78573192-78573192 on Build GRCh38
rs780759567 Chr.4: 78573087-78573087 on Build GRCh38
rs5945 Chr.4: 78573276-78573276 on Build GRCh38
rs188170451 Chr.4: 78573158-78573158 on Build GRCh38
rs376288958 Chr.4: 78573251-78573251 on Build GRCh38
rs943680372 Chr.4: 78573221-78573221 on Build GRCh38
rs755182880 Chr.4: 78573307-78573307 on Build GRCh38
rs766955585 Chr.4: 78573186-78573186 on Build GRCh38
rs777527356 Chr.4: 78573118-78573118 on Build GRCh38
rs148328676 Chr.4: 78573228-78573228 on Build GRCh38
rs753656153 Chr.4: 78573224-78573224 on Build GRCh38
rs111359298 Chr.4: 78573153-78573153 on Build GRCh38
rs184969337 Chr.4: 78573113-78573113 on Build GRCh38
rs532280052 Chr.4: 78573291-78573291 on Build GRCh38
rs769688316 Chr.4: 78573223-78573223 on Build GRCh38

View Full Product Details