Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129637830-129638061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB6
Entrez Gene ID
140459
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
UniGene
Hs.125037
Gene Chromosome Location
Chr.9: 129634604-129642169 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4761476 c.1217A>C p.K406T Chr.9: 129637839-129637839 on Build GRCh38
1105862 c.1074C>T p.I358I Chr.9: 129637982-129637982 on Build GRCh38
6605588 c.1050T>G p.P350P Chr.9: 129638006-129638006 on Build GRCh38
1330698 c.1166C>T p.P389L Chr.9: 129637890-129637890 on Build GRCh38
1105861 c.1165C>T p.P389S Chr.9: 129637891-129637891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750994437 Chr.9: 129638004-129638004 on Build GRCh38
rs200707909 Chr.9: 129637889-129637889 on Build GRCh38
rs1029572669 Chr.9: 129637916-129637916 on Build GRCh38
rs764334444 Chr.9: 129637975-129637975 on Build GRCh38
rs1026649921 Chr.9: 129637985-129637985 on Build GRCh38
rs993902398 Chr.9: 129637984-129637984 on Build GRCh38
rs918251247 Chr.9: 129638001-129638001 on Build GRCh38
rs754198641 Chr.9: 129638018-129638018 on Build GRCh38
rs769783503 Chr.9: 129637957-129637957 on Build GRCh38
rs952521220 Chr.9: 129638006-129638006 on Build GRCh38
rs772763780 Chr.9: 129637874-129637874 on Build GRCh38
rs998050722 Chr.9: 129637915-129637915 on Build GRCh38
rs780392964 Chr.9: 129637921-129637921 on Build GRCh38
rs374183739 Chr.9: 129637952-129637952 on Build GRCh38
rs141198008 Chr.9: 129638026-129638026 on Build GRCh38
rs148649978 Chr.9: 129638017-129638017 on Build GRCh38
rs138458518 Chr.9: 129637931-129637931 on Build GRCh38
rs142387881 Chr.9: 129637963-129637963 on Build GRCh38
rs199686158 Chr.9: 129637905-129637905 on Build GRCh38
rs771015223 Chr.9: 129637942-129637942 on Build GRCh38
rs776601036 Chr.9: 129637890-129637890 on Build GRCh38
rs759843312 Chr.9: 129638027-129638027 on Build GRCh38
rs369236250 Chr.9: 129637991-129637991 on Build GRCh38
rs764140909 Chr.9: 129637906-129637906 on Build GRCh38
rs777189353 Chr.9: 129637937-129637937 on Build GRCh38
rs756761460 Chr.9: 129638028-129638028 on Build GRCh38
rs374558502 Chr.9: 129637870-129637870 on Build GRCh38
rs145897651 Chr.9: 129637850-129637850 on Build GRCh38
rs762815742 Chr.9: 129637924-129637924 on Build GRCh38
rs778475042 Chr.9: 129637857-129637857 on Build GRCh38
rs368489586 Chr.9: 129637914-129637914 on Build GRCh38
rs371398374 Chr.9: 129637917-129637917 on Build GRCh38
rs768314791 Chr.9: 129637854-129637854 on Build GRCh38
rs752799886 Chr.9: 129637899-129637899 on Build GRCh38
rs768013886 Chr.9: 129637996-129637996 on Build GRCh38
rs781625054 Chr.9: 129637918-129637918 on Build GRCh38
rs1043063527 Chr.9: 129637878-129637878 on Build GRCh38
rs768562696 Chr.9: 129637963-129637965 on Build GRCh38
rs746489802 Chr.9: 129637938-129637938 on Build GRCh38
rs774314449 Chr.9: 129637986-129637986 on Build GRCh38
rs747697267 Chr.9: 129637861-129637861 on Build GRCh38
rs534325718 Chr.9: 129637892-129637892 on Build GRCh38
rs759356227 Chr.9: 129637891-129637891 on Build GRCh38
rs985670679 Chr.9: 129637949-129637949 on Build GRCh38
rs750796506 Chr.9: 129637919-129637919 on Build GRCh38
rs776652325 Chr.9: 129637944-129637944 on Build GRCh38
rs1019294578 Chr.9: 129637964-129637964 on Build GRCh38
rs771732219 Chr.9: 129637934-129637934 on Build GRCh38
rs763136780 Chr.9: 129637970-129637970 on Build GRCh38
rs760577422 Chr.9: 129637880-129637880 on Build GRCh38
rs145932765 Chr.9: 129637992-129637992 on Build GRCh38

View Full Product Details