Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149316178-149316433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L1
Entrez Gene ID
134265
Gene Name
actin filament associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.483793
Gene Chromosome Location
Chr.5: 149271838-149343637 on Build GRCh38
Transcripts
NM_001146337.2 NM_001323062.1 NM_152406.3 XM_011537558.1 XM_017009036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
736426 c.1158G>C p.M386I Chr.5: 149316194-149316194 on Build GRCh38
4522340 c.1158G>A p.M386I Chr.5: 149316194-149316194 on Build GRCh38
5522057 c.1244C>T p.T415I Chr.5: 149316280-149316280 on Build GRCh38
351169 c.1225G>T p.D409Y Chr.5: 149316261-149316261 on Build GRCh38
291450 c.1251G>A p.E417E Chr.5: 149316287-149316287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779978586 Chr.5: 149316225-149316225 on Build GRCh38
rs369601847 Chr.5: 149316223-149316223 on Build GRCh38
rs147967184 Chr.5: 149316209-149316209 on Build GRCh38
rs748785548 Chr.5: 149316372-149316372 on Build GRCh38
rs766251670 Chr.5: 149316313-149316313 on Build GRCh38
rs771284582 Chr.5: 149316346-149316346 on Build GRCh38
rs557346448 Chr.5: 149316257-149316257 on Build GRCh38
rs202022315 Chr.5: 149316340-149316340 on Build GRCh38
rs993236873 Chr.5: 149316381-149316381 on Build GRCh38
rs970512661 Chr.5: 149316397-149316397 on Build GRCh38
rs775693791 Chr.5: 149316282-149316282 on Build GRCh38
rs750250368 Chr.5: 149316303-149316303 on Build GRCh38
rs553684604 Chr.5: 149316405-149316405 on Build GRCh38
rs774297897 Chr.5: 149316207-149316207 on Build GRCh38
rs780736714 Chr.5: 149316321-149316321 on Build GRCh38
rs966633558 Chr.5: 149316371-149316371 on Build GRCh38
rs190469835 Chr.5: 149316264-149316264 on Build GRCh38
rs375987421 Chr.5: 149316281-149316281 on Build GRCh38
rs960093339 Chr.5: 149316377-149316377 on Build GRCh38
rs1008960794 Chr.5: 149316277-149316277 on Build GRCh38
rs1035337277 Chr.5: 149316375-149316375 on Build GRCh38
rs929462103 Chr.5: 149316404-149316404 on Build GRCh38
rs150469518 Chr.5: 149316258-149316258 on Build GRCh38
rs1022096445 Chr.5: 149316365-149316365 on Build GRCh38
rs1010663198 Chr.5: 149316369-149316369 on Build GRCh38
rs754464325 Chr.5: 149316243-149316243 on Build GRCh38
rs775783819 Chr.5: 149316350-149316350 on Build GRCh38
rs969223739 Chr.5: 149316389-149316389 on Build GRCh38
rs889988386 Chr.5: 149316330-149316330 on Build GRCh38
rs191310766 Chr.5: 149316218-149316218 on Build GRCh38
rs916032398 Chr.5: 149316388-149316388 on Build GRCh38
rs779112837 Chr.5: 149316348-149316348 on Build GRCh38
rs114156302 Chr.5: 149316253-149316253 on Build GRCh38
rs758830317 Chr.5: 149316211-149316211 on Build GRCh38
rs780854910 Chr.5: 149316244-149316244 on Build GRCh38
rs766902835 Chr.5: 149316206-149316206 on Build GRCh38
rs753962546 Chr.5: 149316240-149316242 on Build GRCh38
rs755734545 Chr.5: 149316271-149316271 on Build GRCh38
rs113355090 Chr.5: 149316262-149316262 on Build GRCh38
rs761039727 Chr.5: 149316286-149316286 on Build GRCh38
rs779750879 Chr.5: 149316349-149316349 on Build GRCh38
rs920740726 Chr.5: 149316402-149316402 on Build GRCh38
rs973545976 Chr.5: 149316400-149316400 on Build GRCh38
rs749634030 Chr.5: 149316342-149316342 on Build GRCh38
rs746515510 Chr.5: 149316237-149316237 on Build GRCh38
rs200946146 Chr.5: 149316222-149316222 on Build GRCh38
rs754840996 Chr.5: 149316319-149316319 on Build GRCh38
rs915965700 Chr.5: 149316214-149316214 on Build GRCh38
rs759561738 Chr.5: 149316272-149316272 on Build GRCh38
rs756369175 Chr.5: 149316215-149316215 on Build GRCh38
rs529146289 Chr.5: 149316210-149316210 on Build GRCh38
rs901387743 Chr.5: 149316361-149316366 on Build GRCh38
rs758155865 Chr.5: 149316311-149316311 on Build GRCh38
rs369729641 Chr.5: 149316332-149316332 on Build GRCh38
rs755766847 Chr.5: 149316333-149316333 on Build GRCh38
rs574140538 Chr.5: 149316318-149316318 on Build GRCh38
rs764409604 Chr.5: 149316287-149316287 on Build GRCh38

View Full Product Details