Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109159224-109159463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP57L1
Entrez Gene ID
285753
Gene Name
centrosomal protein 57 like 1
Gene Aliases
C6orf182, bA487F23.2, cep57R
UniGene
Hs.632616
Gene Chromosome Location
Chr.6: 109095130-109167696 on Build GRCh38
Transcripts
NM_001083535.1 NM_001271852.1 NM_001271853.1 NM_173830.4 XM_005266935.4 XM_005266936.4 XM_005266937.4 XM_005266938.4 XM_006715462.3 XM_011535759.2 XM_011535762.2 XM_017010764.1 XM_017010765.1 XM_017010767.1 XM_017010768.1 XM_017010769.1 XM_017010770.1 XM_017010771.1 XM_017010772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69322 c.1016+1G>A p.? Chr.6: 109159463-109159463 on Build GRCh38
6193350 c.823A>G p.M275V Chr.6: 109159269-109159269 on Build GRCh38
5415135 c.836G>A p.R279H Chr.6: 109159282-109159282 on Build GRCh38
6172210 c.842C>T p.P281L Chr.6: 109159288-109159288 on Build GRCh38
5880384 c.913T>C p.W305R Chr.6: 109159359-109159359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757662191 Chr.6: 109159306-109159306 on Build GRCh38
rs751122664 Chr.6: 109159401-109159401 on Build GRCh38
rs1013871019 Chr.6: 109159421-109159421 on Build GRCh38
rs780793624 Chr.6: 109159335-109159335 on Build GRCh38
rs773151588 Chr.6: 109159340-109159340 on Build GRCh38
rs747740914 Chr.6: 109159432-109159432 on Build GRCh38
rs760646930 Chr.6: 109159354-109159354 on Build GRCh38
rs746670584 Chr.6: 109159319-109159319 on Build GRCh38
rs376132590 Chr.6: 109159251-109159251 on Build GRCh38
rs779451274 Chr.6: 109159394-109159394 on Build GRCh38
rs764390366 Chr.6: 109159288-109159288 on Build GRCh38
rs531072174 Chr.6: 109159313-109159313 on Build GRCh38
rs773129470 Chr.6: 109159245-109159245 on Build GRCh38
rs765639518 Chr.6: 109159386-109159386 on Build GRCh38
rs754301742 Chr.6: 109159291-109159291 on Build GRCh38
rs778636966 Chr.6: 109159392-109159392 on Build GRCh38
rs61743585 Chr.6: 109159339-109159339 on Build GRCh38
rs768405682 Chr.6: 109159322-109159322 on Build GRCh38
rs753368696 Chr.6: 109159249-109159250 on Build GRCh38
rs758897354 Chr.6: 109159389-109159389 on Build GRCh38
rs931807375 Chr.6: 109159356-109159356 on Build GRCh38
rs61743581 Chr.6: 109159359-109159359 on Build GRCh38
rs754105492 Chr.6: 109159357-109159357 on Build GRCh38
rs908976008 Chr.6: 109159374-109159374 on Build GRCh38
rs779626192 Chr.6: 109159312-109159312 on Build GRCh38
rs759432894 Chr.6: 109159360-109159360 on Build GRCh38
rs780881618 Chr.6: 109159431-109159431 on Build GRCh38
rs760914731 Chr.6: 109159282-109159282 on Build GRCh38
rs779548777 Chr.6: 109159314-109159314 on Build GRCh38
rs896126520 Chr.6: 109159402-109159402 on Build GRCh38
rs752869855 Chr.6: 109159274-109159274 on Build GRCh38
rs576180261 Chr.6: 109159377-109159377 on Build GRCh38
rs866403561 Chr.6: 109159324-109159324 on Build GRCh38
rs769585485 Chr.6: 109159438-109159438 on Build GRCh38
rs764414035 Chr.6: 109159355-109159355 on Build GRCh38
rs762886092 Chr.6: 109159341-109159341 on Build GRCh38
rs374194764 Chr.6: 109159427-109159427 on Build GRCh38
rs753485092 Chr.6: 109159342-109159342 on Build GRCh38
rs779549161 Chr.6: 109159361-109159361 on Build GRCh38
rs750969345 Chr.6: 109159387-109159387 on Build GRCh38
rs966019359 Chr.6: 109159252-109159252 on Build GRCh38
rs369162797 Chr.6: 109159395-109159395 on Build GRCh38
rs555480044 Chr.6: 109159351-109159351 on Build GRCh38

View Full Product Details