Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97590164-97590393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf62
Entrez Gene ID
414157
Gene Name
chromosome 10 open reading frame 62
Gene Aliases
bA548K23.1
UniGene
Hs.662302
Gene Chromosome Location
Chr.10: 97589693-97590934 on Build GRCh38
Transcripts
NM_001009997.2 NM_001134670.1 NM_138413.3

Gene Symbol
HOGA1
Entrez Gene ID
112817
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
UniGene
Hs.180346
Gene Chromosome Location
Chr.10: 97584345-97612802 on Build GRCh38
Transcripts
NM_001009997.2 NM_001134670.1 NM_138413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414837 c.490A>T p.N164Y Chr.10: 97590387-97590387 on Build GRCh38
4971453 c.366C>A p.A122A Chr.10: 97590263-97590263 on Build GRCh38
1638715 c.493C>A p.Q165K Chr.10: 97590390-97590390 on Build GRCh38
922046 c.302C>A p.S101Y Chr.10: 97590199-97590199 on Build GRCh38
3808133 c.345A>T p.K115N Chr.10: 97590242-97590242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748010979 Chr.10: 97590285-97590285 on Build GRCh38
rs755053963 Chr.10: 97590333-97590333 on Build GRCh38
rs758539967 Chr.10: 97590261-97590261 on Build GRCh38
rs754349047 Chr.10: 97590254-97590254 on Build GRCh38
rs757729928 Chr.10: 97590257-97590257 on Build GRCh38
rs772854255 Chr.10: 97590183-97590183 on Build GRCh38
rs780159949 Chr.10: 97590329-97590329 on Build GRCh38
rs368370225 Chr.10: 97590243-97590243 on Build GRCh38
rs781188795 Chr.10: 97590335-97590335 on Build GRCh38
rs777584853 Chr.10: 97590344-97590344 on Build GRCh38
rs76383275 Chr.10: 97590346-97590346 on Build GRCh38
rs750719654 Chr.10: 97590325-97590325 on Build GRCh38
rs762732537 Chr.10: 97590298-97590298 on Build GRCh38
rs754404049 Chr.10: 97590312-97590312 on Build GRCh38
rs373892055 Chr.10: 97590204-97590204 on Build GRCh38
rs7093840 Chr.10: 97590260-97590260 on Build GRCh38
rs7093643 Chr.10: 97590371-97590371 on Build GRCh38
rs199811878 Chr.10: 97590348-97590348 on Build GRCh38
rs751762663 Chr.10: 97590332-97590332 on Build GRCh38
rs775259178 Chr.10: 97590354-97590354 on Build GRCh38
rs375555828 Chr.10: 97590361-97590361 on Build GRCh38
rs150921567 Chr.10: 97590317-97590317 on Build GRCh38
rs762455396 Chr.10: 97590192-97590192 on Build GRCh38
rs768730543 Chr.10: 97590273-97590273 on Build GRCh38
rs999178434 Chr.10: 97590258-97590258 on Build GRCh38
rs565007098 Chr.10: 97590266-97590266 on Build GRCh38
rs772977970 Chr.10: 97590288-97590288 on Build GRCh38
rs770460210 Chr.10: 97590305-97590305 on Build GRCh38
rs764677438 Chr.10: 97590248-97590248 on Build GRCh38
rs769587708 Chr.10: 97590287-97590287 on Build GRCh38
rs747034275 Chr.10: 97590272-97590272 on Build GRCh38
rs748141701 Chr.10: 97590339-97590339 on Build GRCh38
rs371888091 Chr.10: 97590308-97590308 on Build GRCh38
rs773907690 Chr.10: 97590359-97590359 on Build GRCh38
rs1012482351 Chr.10: 97590302-97590302 on Build GRCh38
rs769717721 Chr.10: 97590340-97590340 on Build GRCh38
rs1012895434 Chr.10: 97590307-97590307 on Build GRCh38
rs781135280 Chr.10: 97590276-97590276 on Build GRCh38
rs758692283 Chr.10: 97590326-97590326 on Build GRCh38
rs765971580 Chr.10: 97590208-97590208 on Build GRCh38
rs142514598 Chr.10: 97590313-97590313 on Build GRCh38
rs147809962 Chr.10: 97590238-97590238 on Build GRCh38
rs759164296 Chr.10: 97590365-97590365 on Build GRCh38

View Full Product Details