Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29061394-29061655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200211 c.1288T>C p.C430R Chr.21: 29061414-29061414 on Build GRCh38
6219025 c.1274C>T p.S425L Chr.21: 29061506-29061506 on Build GRCh38
5628307 c.1283A>T p.E428V Chr.21: 29061497-29061497 on Build GRCh38
6401448 c.1232G>T p.G411V Chr.21: 29061548-29061548 on Build GRCh38
5644287 c.1306T>A p.Y436N Chr.21: 29061396-29061396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962475513 Chr.21: 29061453-29061453 on Build GRCh38
rs116595826 Chr.21: 29061614-29061614 on Build GRCh38
rs567041193 Chr.21: 29061587-29061588 on Build GRCh38
rs747713434 Chr.21: 29061588-29061588 on Build GRCh38
rs772638602 Chr.21: 29061573-29061573 on Build GRCh38
rs115160321 Chr.21: 29061470-29061470 on Build GRCh38
rs745871687 Chr.21: 29061432-29061432 on Build GRCh38
rs944421784 Chr.21: 29061424-29061424 on Build GRCh38
rs762752300 Chr.21: 29061612-29061612 on Build GRCh38
rs970191064 Chr.21: 29061528-29061528 on Build GRCh38
rs8129954 Chr.21: 29061555-29061555 on Build GRCh38
rs537922342 Chr.21: 29061513-29061513 on Build GRCh38
rs569435050 Chr.21: 29061489-29061489 on Build GRCh38
rs762070047 Chr.21: 29061590-29061590 on Build GRCh38
rs368274683 Chr.21: 29061441-29061441 on Build GRCh38
rs1005812210 Chr.21: 29061471-29061471 on Build GRCh38
rs1024797138 Chr.21: 29061514-29061514 on Build GRCh38
rs776360584 Chr.21: 29061449-29061449 on Build GRCh38
rs575020223 Chr.21: 29061584-29061584 on Build GRCh38
rs375555917 Chr.21: 29061482-29061482 on Build GRCh38
rs1044886890 Chr.21: 29061564-29061564 on Build GRCh38
rs765056082 Chr.21: 29061527-29061527 on Build GRCh38
rs548773037 Chr.21: 29061511-29061511 on Build GRCh38
rs746264218 Chr.21: 29061571-29061571 on Build GRCh38
rs932801778 Chr.21: 29061455-29061455 on Build GRCh38
rs371049669 Chr.21: 29061467-29061467 on Build GRCh38
rs779586870 Chr.21: 29061439-29061439 on Build GRCh38
rs772887141 Chr.21: 29061588-29061589 on Build GRCh38
rs750977190 Chr.21: 29061546-29061546 on Build GRCh38
rs776685936 Chr.21: 29061586-29061586 on Build GRCh38
rs779973549 Chr.21: 29061541-29061541 on Build GRCh38
rs376306326 Chr.21: 29061433-29061433 on Build GRCh38
rs750337571 Chr.21: 29061537-29061537 on Build GRCh38
rs760172230 Chr.21: 29061468-29061468 on Build GRCh38
rs554139712 Chr.21: 29061570-29061571 on Build GRCh38
rs765466113 Chr.21: 29061592-29061592 on Build GRCh38
rs367948039 Chr.21: 29061476-29061476 on Build GRCh38
rs778655265 Chr.21: 29061579-29061580 on Build GRCh38
rs948012155 Chr.21: 29061560-29061560 on Build GRCh38
rs752852837 Chr.21: 29061550-29061550 on Build GRCh38
rs760655089 Chr.21: 29061582-29061582 on Build GRCh38
rs776073376 Chr.21: 29061578-29061578 on Build GRCh38
rs1016886043 Chr.21: 29061435-29061435 on Build GRCh38
rs566821834 Chr.21: 29061461-29061461 on Build GRCh38
rs750153549 Chr.21: 29061598-29061598 on Build GRCh38
rs373037770 Chr.21: 29061552-29061552 on Build GRCh38
rs779375522 Chr.21: 29061561-29061561 on Build GRCh38
rs372173315 Chr.21: 29061479-29061479 on Build GRCh38
rs369493335 Chr.21: 29061503-29061503 on Build GRCh38
rs768128292 Chr.21: 29061447-29061447 on Build GRCh38

View Full Product Details