Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74592459-74592714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf77
Entrez Gene ID
146723
Gene Name
chromosome 17 open reading frame 77
Gene Aliases
-
UniGene
Hs.350775
Gene Chromosome Location
Chr.17: 74584679-74594209 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148004 c.549C>A p.C183* Chr.17: 74592595-74592595 on Build GRCh38
437323 c.435G>A p.T145T Chr.17: 74592481-74592481 on Build GRCh38
437324 c.476G>A p.G159E Chr.17: 74592522-74592522 on Build GRCh38
6302657 c.524C>A p.T175K Chr.17: 74592570-74592570 on Build GRCh38
4069376 c.524C>T p.T175M Chr.17: 74592570-74592570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774561890 Chr.17: 74592562-74592562 on Build GRCh38
rs1052701074 Chr.17: 74592477-74592477 on Build GRCh38
rs754359220 Chr.17: 74592515-74592515 on Build GRCh38
rs58253413 Chr.17: 74592639-74592639 on Build GRCh38
rs773729422 Chr.17: 74592546-74592546 on Build GRCh38
rs767633084 Chr.17: 74592491-74592491 on Build GRCh38
rs764503796 Chr.17: 74592593-74592593 on Build GRCh38
rs751394899 Chr.17: 74592683-74592683 on Build GRCh38
rs775546074 Chr.17: 74592577-74592577 on Build GRCh38
rs191930489 Chr.17: 74592689-74592689 on Build GRCh38
rs768145771 Chr.17: 74592481-74592481 on Build GRCh38
rs758137297 Chr.17: 74592531-74592531 on Build GRCh38
rs267605041 Chr.17: 74592537-74592537 on Build GRCh38
rs757822480 Chr.17: 74592626-74592626 on Build GRCh38
rs369351414 Chr.17: 74592679-74592679 on Build GRCh38
rs374939995 Chr.17: 74592621-74592621 on Build GRCh38
rs113009897 Chr.17: 74592490-74592490 on Build GRCh38
rs1022042874 Chr.17: 74592486-74592486 on Build GRCh38
rs566495980 Chr.17: 74592532-74592532 on Build GRCh38
rs780904053 Chr.17: 74592645-74592645 on Build GRCh38
rs752488459 Chr.17: 74592654-74592654 on Build GRCh38
rs761102703 Chr.17: 74592496-74592496 on Build GRCh38
rs142909208 Chr.17: 74592596-74592596 on Build GRCh38
rs749407544 Chr.17: 74592480-74592480 on Build GRCh38
rs372731087 Chr.17: 74592571-74592571 on Build GRCh38
rs369204239 Chr.17: 74592533-74592533 on Build GRCh38
rs764748120 Chr.17: 74592522-74592522 on Build GRCh38
rs963927528 Chr.17: 74592487-74592487 on Build GRCh38
rs145962818 Chr.17: 74592485-74592485 on Build GRCh38
rs761154100 Chr.17: 74592592-74592592 on Build GRCh38
rs138887403 Chr.17: 74592530-74592530 on Build GRCh38
rs996971299 Chr.17: 74592509-74592509 on Build GRCh38
rs745925063 Chr.17: 74592570-74592570 on Build GRCh38
rs916057916 Chr.17: 74592576-74592576 on Build GRCh38
rs545652 Chr.17: 74592667-74592667 on Build GRCh38
rs148306662 Chr.17: 74592662-74592662 on Build GRCh38
rs766092775 Chr.17: 74592632-74592632 on Build GRCh38
rs754734146 Chr.17: 74592544-74592544 on Build GRCh38
rs571801130 Chr.17: 74592494-74592494 on Build GRCh38
rs766361173 Chr.17: 74592505-74592506 on Build GRCh38
rs200171656 Chr.17: 74592644-74592644 on Build GRCh38
rs539574024 Chr.17: 74592518-74592518 on Build GRCh38
rs764740464 Chr.17: 74592514-74592514 on Build GRCh38
rs868622640 Chr.17: 74592542-74592542 on Build GRCh38
rs781136951 Chr.17: 74592545-74592545 on Build GRCh38
rs765657104 Chr.17: 74592609-74592609 on Build GRCh38
rs200986630 Chr.17: 74592601-74592601 on Build GRCh38
rs772007436 Chr.17: 74592676-74592676 on Build GRCh38
rs770924155 Chr.17: 74592552-74592552 on Build GRCh38
rs748025471 Chr.17: 74592548-74592548 on Build GRCh38

View Full Product Details