Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25389963-25390210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7167618 c.556G>C p.E186Q Chr.4: 25390176-25390176 on Build GRCh38
673928 c.529G>A p.V177I Chr.4: 25390149-25390149 on Build GRCh38
5866231 c.566C>T p.A189V Chr.4: 25390186-25390186 on Build GRCh38
733655 c.556G>T p.E186* Chr.4: 25390176-25390176 on Build GRCh38
6420619 c.559C>A p.L187I Chr.4: 25390179-25390179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007059061 Chr.4: 25390080-25390080 on Build GRCh38
rs549611082 Chr.4: 25390070-25390070 on Build GRCh38
rs760426798 Chr.4: 25390106-25390106 on Build GRCh38
rs988848095 Chr.4: 25390163-25390163 on Build GRCh38
rs777518592 Chr.4: 25390178-25390178 on Build GRCh38
rs184720412 Chr.4: 25390113-25390113 on Build GRCh38
rs1004061270 Chr.4: 25390139-25390139 on Build GRCh38
rs764282053 Chr.4: 25390100-25390100 on Build GRCh38
rs111628527 Chr.4: 25390034-25390034 on Build GRCh38
rs752115448 Chr.4: 25390149-25390149 on Build GRCh38
rs900585533 Chr.4: 25390053-25390053 on Build GRCh38
rs754859496 Chr.4: 25390131-25390131 on Build GRCh38
rs962667806 Chr.4: 25390154-25390154 on Build GRCh38
rs149911152 Chr.4: 25390148-25390148 on Build GRCh38
rs535218219 Chr.4: 25390173-25390173 on Build GRCh38
rs753752934 Chr.4: 25390119-25390119 on Build GRCh38
rs879467575 Chr.4: 25390008-25390008 on Build GRCh38
rs374391579 Chr.4: 25390086-25390086 on Build GRCh38
rs773877660 Chr.4: 25390076-25390076 on Build GRCh38
rs370102686 Chr.4: 25390085-25390085 on Build GRCh38
rs1026720149 Chr.4: 25390006-25390006 on Build GRCh38
rs889990356 Chr.4: 25390064-25390064 on Build GRCh38

View Full Product Details