Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79729145-79729412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMYA5
Entrez Gene ID
202333
Gene Name
cardiomyopathy associated 5
Gene Aliases
C5orf10, SPRYD2, TRIM76
UniGene
Hs.482625
Gene Chromosome Location
Chr.5: 79689339-79800226 on Build GRCh38
Transcripts
NM_153610.4 XM_017009212.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618089 c.607C>T p.P203S Chr.5: 79729372-79729372 on Build GRCh38
5868845 c.506C>T p.P169L Chr.5: 79729271-79729271 on Build GRCh38
3761335 c.569A>G p.D190G Chr.5: 79729334-79729334 on Build GRCh38
4408333 c.469G>T p.E157* Chr.5: 79729234-79729234 on Build GRCh38
5603222 c.609G>A p.P203P Chr.5: 79729374-79729374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191385245 Chr.5: 79729379-79729379 on Build GRCh38
rs181719440 Chr.5: 79729232-79729232 on Build GRCh38
rs371426208 Chr.5: 79729218-79729218 on Build GRCh38
rs781301181 Chr.5: 79729249-79729249 on Build GRCh38
rs768454519 Chr.5: 79729361-79729361 on Build GRCh38
rs780017437 Chr.5: 79729335-79729335 on Build GRCh38
rs886403941 Chr.5: 79729173-79729173 on Build GRCh38
rs765412906 Chr.5: 79729316-79729316 on Build GRCh38
rs768242479 Chr.5: 79729270-79729270 on Build GRCh38
rs769716466 Chr.5: 79729254-79729254 on Build GRCh38
rs72635618 Chr.5: 79729262-79729262 on Build GRCh38
rs771571752 Chr.5: 79729374-79729374 on Build GRCh38
rs764974849 Chr.5: 79729241-79729242 on Build GRCh38
rs753783340 Chr.5: 79729185-79729186 on Build GRCh38
rs371922369 Chr.5: 79729172-79729172 on Build GRCh38
rs939604414 Chr.5: 79729342-79729342 on Build GRCh38
rs1030666400 Chr.5: 79729289-79729289 on Build GRCh38
rs766668920 Chr.5: 79729224-79729224 on Build GRCh38
rs186943498 Chr.5: 79729287-79729287 on Build GRCh38
rs779177993 Chr.5: 79729239-79729239 on Build GRCh38
rs139186257 Chr.5: 79729298-79729298 on Build GRCh38
rs376859315 Chr.5: 79729182-79729182 on Build GRCh38
rs778448654 Chr.5: 79729368-79729368 on Build GRCh38
rs755216051 Chr.5: 79729237-79729237 on Build GRCh38
rs777935616 Chr.5: 79729327-79729327 on Build GRCh38
rs554686825 Chr.5: 79729223-79729223 on Build GRCh38
rs771386167 Chr.5: 79729381-79729381 on Build GRCh38
rs775297395 Chr.5: 79729195-79729195 on Build GRCh38
rs531983065 Chr.5: 79729355-79729355 on Build GRCh38
rs745744183 Chr.5: 79729252-79729252 on Build GRCh38
rs761669476 Chr.5: 79729230-79729230 on Build GRCh38
rs930718528 Chr.5: 79729206-79729206 on Build GRCh38
rs758566227 Chr.5: 79729325-79729325 on Build GRCh38
rs201453503 Chr.5: 79729311-79729311 on Build GRCh38
rs570397324 Chr.5: 79729183-79729183 on Build GRCh38
rs188600563 Chr.5: 79729190-79729190 on Build GRCh38
rs750886227 Chr.5: 79729221-79729221 on Build GRCh38
rs752763418 Chr.5: 79729242-79729242 on Build GRCh38
rs373060111 Chr.5: 79729260-79729260 on Build GRCh38
rs987154466 Chr.5: 79729293-79729293 on Build GRCh38
rs10942901 Chr.5: 79729334-79729334 on Build GRCh38
rs904060688 Chr.5: 79729238-79729238 on Build GRCh38
rs752916916 Chr.5: 79729321-79729321 on Build GRCh38
rs62621858 Chr.5: 79729205-79729205 on Build GRCh38
rs373527728 Chr.5: 79729301-79729301 on Build GRCh38
rs542874952 Chr.5: 79729179-79729179 on Build GRCh38
rs751686303 Chr.5: 79729331-79729331 on Build GRCh38
rs762447448 Chr.5: 79729202-79729202 on Build GRCh38
rs750187197 Chr.5: 79729302-79729302 on Build GRCh38
rs764073265 Chr.5: 79729172-79729173 on Build GRCh38
rs747738648 Chr.5: 79729373-79729373 on Build GRCh38
rs555997523 Chr.5: 79729204-79729204 on Build GRCh38
rs547053590 Chr.5: 79729174-79729174 on Build GRCh38
rs927998636 Chr.5: 79729184-79729184 on Build GRCh38
rs749076588 Chr.5: 79729353-79729353 on Build GRCh38
rs939322701 Chr.5: 79729186-79729186 on Build GRCh38
rs758513257 Chr.5: 79729245-79729245 on Build GRCh38
rs6895605 Chr.5: 79729290-79729290 on Build GRCh38
rs754207328 Chr.5: 79729309-79729309 on Build GRCh38
rs766860966 Chr.5: 79729302-79729302 on Build GRCh38
rs761363412 Chr.5: 79729210-79729210 on Build GRCh38

View Full Product Details