Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153905714-153905977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR2
Entrez Gene ID
554
Gene Name
arginine vasopressin receptor 2
Gene Aliases
ADHR, DI1, DIR, DIR3, NDI, V2R
UniGene
Hs.567240
Gene Chromosome Location
Chr.X: 153902531-153907166 on Build GRCh38
Transcripts
NM_000054.4 NM_001146151.1 NR_027419.1 XM_006724828.3

SNP Detail

SNP ID Chromosome Location
rs905300222 Chr.X: 153905951-153905951 on Build GRCh38
rs782453202 Chr.X: 153905767-153905767 on Build GRCh38
rs782511391 Chr.X: 153905732-153905732 on Build GRCh38
rs782571215 Chr.X: 153905936-153905936 on Build GRCh38
rs104894756 Chr.X: 153905916-153905916 on Build GRCh38
rs796052096 Chr.X: 153905894-153905894 on Build GRCh38
rs782077927 Chr.X: 153905874-153905874 on Build GRCh38
rs201053959 Chr.X: 153905823-153905823 on Build GRCh38
rs782549298 Chr.X: 153905907-153905907 on Build GRCh38
rs104894747 Chr.X: 153905901-153905901 on Build GRCh38
rs782224593 Chr.X: 153905957-153905957 on Build GRCh38
rs28935496 Chr.X: 153905843-153905843 on Build GRCh38
rs104894760 Chr.X: 153905816-153905816 on Build GRCh38
rs146238429 Chr.X: 153905934-153905934 on Build GRCh38
rs781808434 Chr.X: 153905812-153905812 on Build GRCh38
rs370880911 Chr.X: 153905758-153905758 on Build GRCh38
rs782151840 Chr.X: 153905848-153905848 on Build GRCh38
rs104894761 Chr.X: 153905915-153905915 on Build GRCh38
rs782616366 Chr.X: 153905923-153905923 on Build GRCh38
rs193922114 Chr.X: 153905930-153905930 on Build GRCh38
rs104894758 Chr.X: 153905819-153905819 on Build GRCh38
rs5200 Chr.X: 153905946-153905946 on Build GRCh38
rs893680024 Chr.X: 153905929-153905929 on Build GRCh38
rs782202028 Chr.X: 153905821-153905821 on Build GRCh38
rs143055328 Chr.X: 153905947-153905947 on Build GRCh38
rs782521961 Chr.X: 153905806-153905806 on Build GRCh38
rs373990379 Chr.X: 153905817-153905817 on Build GRCh38
rs782096959 Chr.X: 153905922-153905922 on Build GRCh38
rs374561268 Chr.X: 153905952-153905952 on Build GRCh38
rs782789114 Chr.X: 153905908-153905908 on Build GRCh38
rs193922113 Chr.X: 153905796-153905796 on Build GRCh38
rs1057518723 Chr.X: 153905841-153905841 on Build GRCh38
rs782038329 Chr.X: 153905846-153905846 on Build GRCh38
rs781950164 Chr.X: 153905889-153905889 on Build GRCh38
rs782026554 Chr.X: 153905742-153905742 on Build GRCh38
rs878853079 Chr.X: 153905898-153905898 on Build GRCh38
rs782291586 Chr.X: 153905795-153905795 on Build GRCh38
rs370616859 Chr.X: 153905891-153905891 on Build GRCh38
rs781956131 Chr.X: 153905818-153905818 on Build GRCh38
rs104894754 Chr.X: 153905759-153905759 on Build GRCh38
rs782753911 Chr.X: 153905849-153905849 on Build GRCh38
rs782323704 Chr.X: 153905865-153905865 on Build GRCh38
rs782565222 Chr.X: 153905776-153905776 on Build GRCh38

View Full Product Details