Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78586492-78586755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
701725 c.294G>T p.W98C Chr.15: 78586680-78586680 on Build GRCh38
5849860 c.276_277delAA p.L92fs*21 Chr.15: 78586662-78586663 on Build GRCh38
965385 c.262G>A p.E88K Chr.15: 78586648-78586648 on Build GRCh38
5670344 c.290T>A p.V97D Chr.15: 78586676-78586676 on Build GRCh38
6628728 c.288C>T p.N96N Chr.15: 78586674-78586674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771582989 Chr.15: 78586710-78586710 on Build GRCh38
rs200704018 Chr.15: 78586623-78586623 on Build GRCh38
rs78060596 Chr.15: 78586545-78586545 on Build GRCh38
rs199649840 Chr.15: 78586664-78586664 on Build GRCh38
rs372734191 Chr.15: 78586720-78586720 on Build GRCh38
rs55982512 Chr.15: 78586716-78586716 on Build GRCh38
rs369401295 Chr.15: 78586615-78586615 on Build GRCh38
rs1034987451 Chr.15: 78586593-78586593 on Build GRCh38
rs199753114 Chr.15: 78586711-78586711 on Build GRCh38
rs776348268 Chr.15: 78586712-78586712 on Build GRCh38
rs757754067 Chr.15: 78586667-78586667 on Build GRCh38
rs75600623 Chr.15: 78586674-78586674 on Build GRCh38
rs573456429 Chr.15: 78586577-78586577 on Build GRCh38
rs749045535 Chr.15: 78586634-78586635 on Build GRCh38
rs935110324 Chr.15: 78586561-78586561 on Build GRCh38
rs770864582 Chr.15: 78586658-78586661 on Build GRCh38
rs558347927 Chr.15: 78586543-78586543 on Build GRCh38
rs372092068 Chr.15: 78586594-78586594 on Build GRCh38
rs200691818 Chr.15: 78586670-78586670 on Build GRCh38
rs778510395 Chr.15: 78586690-78586690 on Build GRCh38
rs555893661 Chr.15: 78586680-78586680 on Build GRCh38
rs1014744827 Chr.15: 78586687-78586687 on Build GRCh38
rs200806416 Chr.15: 78586719-78586719 on Build GRCh38
rs534314741 Chr.15: 78586579-78586579 on Build GRCh38
rs201183620 Chr.15: 78586644-78586644 on Build GRCh38
rs768366429 Chr.15: 78586688-78586688 on Build GRCh38
rs747574001 Chr.15: 78586698-78586698 on Build GRCh38
rs775748132 Chr.15: 78586722-78586722 on Build GRCh38
rs1051314064 Chr.15: 78586613-78586613 on Build GRCh38
rs1041887284 Chr.15: 78586551-78586551 on Build GRCh38
rs758973811 Chr.15: 78586610-78586610 on Build GRCh38
rs924918084 Chr.15: 78586552-78586552 on Build GRCh38
rs907388147 Chr.15: 78586573-78586573 on Build GRCh38
rs1027287255 Chr.15: 78586629-78586629 on Build GRCh38
rs777570565 Chr.15: 78586629-78586629 on Build GRCh38
rs966240090 Chr.15: 78586699-78586699 on Build GRCh38
rs56257539 Chr.15: 78586626-78586626 on Build GRCh38
rs148722844 Chr.15: 78586675-78586675 on Build GRCh38

View Full Product Details