Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196515047-196515318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO1
Entrez Gene ID
255798
Gene Name
single-pass membrane protein with coiled-coil domains 1
Gene Aliases
C3orf43
UniGene
Hs.631933
Gene Chromosome Location
Chr.3: 196506877-196515366 on Build GRCh38
Transcripts
NM_001077657.2 NM_001320473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263806 c.41C>T p.A14V Chr.3: 196515169-196515169 on Build GRCh38
4426121 c.50+1G>T p.? Chr.3: 196515159-196515159 on Build GRCh38
1242071 c.20C>T p.T7I Chr.3: 196515190-196515190 on Build GRCh38
5733766 c.2T>A p.M1K Chr.3: 196515208-196515208 on Build GRCh38
6545160 c.45G>A p.M15I Chr.3: 196515165-196515165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757893839 Chr.3: 196515118-196515118 on Build GRCh38
rs757818277 Chr.3: 196515237-196515246 on Build GRCh38
rs746370295 Chr.3: 196515246-196515246 on Build GRCh38
rs749897707 Chr.3: 196515227-196515231 on Build GRCh38
rs377153433 Chr.3: 196515196-196515196 on Build GRCh38
rs1032499009 Chr.3: 196515238-196515238 on Build GRCh38
rs1008821784 Chr.3: 196515259-196515259 on Build GRCh38
rs779669988 Chr.3: 196515247-196515247 on Build GRCh38
rs370274768 Chr.3: 196515213-196515213 on Build GRCh38
rs757132357 Chr.3: 196515133-196515134 on Build GRCh38
rs369522412 Chr.3: 196515126-196515126 on Build GRCh38
rs551373022 Chr.3: 196515137-196515137 on Build GRCh38
rs931078298 Chr.3: 196515189-196515189 on Build GRCh38
rs757210031 Chr.3: 196515099-196515099 on Build GRCh38
rs768450323 Chr.3: 196515162-196515162 on Build GRCh38
rs749761499 Chr.3: 196515130-196515130 on Build GRCh38
rs887110942 Chr.3: 196515132-196515132 on Build GRCh38
rs920530633 Chr.3: 196515085-196515085 on Build GRCh38
rs760519145 Chr.3: 196515088-196515088 on Build GRCh38
rs772431949 Chr.3: 196515107-196515107 on Build GRCh38
rs778241143 Chr.3: 196515225-196515225 on Build GRCh38
rs771593902 Chr.3: 196515131-196515131 on Build GRCh38
rs746441610 Chr.3: 196515241-196515241 on Build GRCh38
rs776442663 Chr.3: 196515164-196515164 on Build GRCh38
rs751163175 Chr.3: 196515209-196515209 on Build GRCh38
rs566815688 Chr.3: 196515289-196515289 on Build GRCh38
rs560668637 Chr.3: 196515122-196515123 on Build GRCh38
rs772132974 Chr.3: 196515252-196515252 on Build GRCh38
rs533125905 Chr.3: 196515094-196515094 on Build GRCh38
rs1013560702 Chr.3: 196515133-196515133 on Build GRCh38
rs779743415 Chr.3: 196515120-196515120 on Build GRCh38
rs762519916 Chr.3: 196515193-196515193 on Build GRCh38
rs201500241 Chr.3: 196515190-196515190 on Build GRCh38
rs182371322 Chr.3: 196515188-196515188 on Build GRCh38
rs754234737 Chr.3: 196515228-196515228 on Build GRCh38
rs765135565 Chr.3: 196515184-196515184 on Build GRCh38
rs753718782 Chr.3: 196515097-196515097 on Build GRCh38
rs779615648 Chr.3: 196515135-196515135 on Build GRCh38
rs761638721 Chr.3: 196515183-196515183 on Build GRCh38
rs375458427 Chr.3: 196515121-196515121 on Build GRCh38
rs779562647 Chr.3: 196515237-196515237 on Build GRCh38
rs775636320 Chr.3: 196515083-196515083 on Build GRCh38
rs551496314 Chr.3: 196515251-196515252 on Build GRCh38
rs761017956 Chr.3: 196515107-196515108 on Build GRCh38
rs745932403 Chr.3: 196515141-196515141 on Build GRCh38
rs772113824 Chr.3: 196515143-196515143 on Build GRCh38
rs372386896 Chr.3: 196515232-196515232 on Build GRCh38
rs556111938 Chr.3: 196515290-196515290 on Build GRCh38
rs775719353 Chr.3: 196515159-196515159 on Build GRCh38
rs753636025 Chr.3: 196515131-196515133 on Build GRCh38
rs764688684 Chr.3: 196515104-196515104 on Build GRCh38
rs749901849 Chr.3: 196515106-196515106 on Build GRCh38
rs866126470 Chr.3: 196515073-196515073 on Build GRCh38
rs775919393 Chr.3: 196515124-196515124 on Build GRCh38
rs763802295 Chr.3: 196515090-196515090 on Build GRCh38

View Full Product Details