Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65536335-65536584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXIN2
Entrez Gene ID
8313
Gene Name
axin 2
Gene Aliases
AXIL, ODCRCS
UniGene
Hs.156527
Gene Chromosome Location
Chr.17: 65528563-65561622 on Build GRCh38
Transcripts
NM_004655.3 XM_011525319.2 XM_011525320.1 XM_011525321.2 XM_017025192.1 XM_017025193.1 XM_017025194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5945894 c.2024G>A p.R675H Chr.17: 65536437-65536437 on Build GRCh38
6943376 c.1990G>A p.G664R Chr.17: 65536471-65536471 on Build GRCh38
4476499 c.2072C>T p.P691L Chr.17: 65536389-65536389 on Build GRCh38
4925502 c.1984C>T p.L662L Chr.17: 65536477-65536477 on Build GRCh38
707610 c.2023C>T p.R675C Chr.17: 65536438-65536438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368798367 Chr.17: 65536428-65536428 on Build GRCh38
rs140510381 Chr.17: 65536378-65536378 on Build GRCh38
rs576688801 Chr.17: 65536443-65536443 on Build GRCh38
rs776430866 Chr.17: 65536469-65536469 on Build GRCh38
rs771109399 Chr.17: 65536401-65536401 on Build GRCh38
rs730881401 Chr.17: 65536437-65536437 on Build GRCh38
rs551098654 Chr.17: 65536382-65536382 on Build GRCh38
rs900176493 Chr.17: 65536421-65536421 on Build GRCh38
rs772202125 Chr.17: 65536467-65536467 on Build GRCh38
rs878854724 Chr.17: 65536461-65536461 on Build GRCh38
rs74006838 Chr.17: 65536509-65536509 on Build GRCh38
rs968390172 Chr.17: 65536545-65536545 on Build GRCh38
rs142670753 Chr.17: 65536486-65536486 on Build GRCh38
rs767111161 Chr.17: 65536498-65536498 on Build GRCh38
rs199719878 Chr.17: 65536512-65536512 on Build GRCh38
rs565565172 Chr.17: 65536353-65536353 on Build GRCh38
rs917901663 Chr.17: 65536507-65536507 on Build GRCh38
rs754480214 Chr.17: 65536490-65536490 on Build GRCh38
rs367725839 Chr.17: 65536395-65536395 on Build GRCh38
rs1025003994 Chr.17: 65536530-65536530 on Build GRCh38
rs138287857 Chr.17: 65536410-65536410 on Build GRCh38
rs779214199 Chr.17: 65536518-65536518 on Build GRCh38
rs979932202 Chr.17: 65536477-65536477 on Build GRCh38
rs1047911557 Chr.17: 65536406-65536406 on Build GRCh38
rs770199204 Chr.17: 65536393-65536393 on Build GRCh38
rs756419208 Chr.17: 65536454-65536454 on Build GRCh38
rs151279728 Chr.17: 65536437-65536448 on Build GRCh38
rs148419444 Chr.17: 65536513-65536513 on Build GRCh38
rs377642903 Chr.17: 65536499-65536499 on Build GRCh38
rs730882193 Chr.17: 65536472-65536472 on Build GRCh38
rs376550544 Chr.17: 65536521-65536521 on Build GRCh38
rs765149006 Chr.17: 65536438-65536438 on Build GRCh38
rs778376993 Chr.17: 65536374-65536374 on Build GRCh38
rs747355609 Chr.17: 65536466-65536467 on Build GRCh38
rs781738256 Chr.17: 65536388-65536388 on Build GRCh38
rs761870581 Chr.17: 65536473-65536473 on Build GRCh38
rs556551269 Chr.17: 65536439-65536439 on Build GRCh38
rs111650787 Chr.17: 65536408-65536408 on Build GRCh38
rs781484593 Chr.17: 65536451-65536451 on Build GRCh38
rs142476324 Chr.17: 65536476-65536476 on Build GRCh38
rs749487037 Chr.17: 65536456-65536456 on Build GRCh38
rs372086777 Chr.17: 65536508-65536508 on Build GRCh38
rs267606674 Chr.17: 65536467-65536467 on Build GRCh38
rs757100957 Chr.17: 65536356-65536356 on Build GRCh38
rs35415678 Chr.17: 65536399-65536399 on Build GRCh38
rs774732785 Chr.17: 65536459-65536459 on Build GRCh38
rs200573256 Chr.17: 65536409-65536409 on Build GRCh38
rs978837790 Chr.17: 65536555-65536555 on Build GRCh38
rs755519590 Chr.17: 65536383-65536383 on Build GRCh38
rs770763142 Chr.17: 65536474-65536474 on Build GRCh38
rs754506970 Chr.17: 65536450-65536450 on Build GRCh38
rs730881405 Chr.17: 65536527-65536527 on Build GRCh38
rs1030272364 Chr.17: 65536434-65536434 on Build GRCh38
rs748632114 Chr.17: 65536427-65536427 on Build GRCh38
rs730881400 Chr.17: 65536485-65536485 on Build GRCh38
rs765845684 Chr.17: 65536449-65536449 on Build GRCh38
rs144092307 Chr.17: 65536457-65536457 on Build GRCh38
rs911977110 Chr.17: 65536420-65536420 on Build GRCh38
rs780233103 Chr.17: 65536532-65536532 on Build GRCh38
rs748005374 Chr.17: 65536534-65536534 on Build GRCh38
rs748402111 Chr.17: 65536391-65536391 on Build GRCh38
rs376242268 Chr.17: 65536460-65536460 on Build GRCh38
rs751216221 Chr.17: 65536487-65536487 on Build GRCh38
rs773157765 Chr.17: 65536554-65536554 on Build GRCh38
rs748754551 Chr.17: 65536452-65536452 on Build GRCh38
rs750097494 Chr.17: 65536376-65536376 on Build GRCh38
rs121908568 Chr.17: 65536495-65536495 on Build GRCh38
rs199724028 Chr.17: 65536478-65536478 on Build GRCh38

View Full Product Details