Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69153770-69154031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298869 c.3803G>A p.S1268N Chr.17: 69153993-69153993 on Build GRCh38
390527 c.3787-1G>C p.? Chr.17: 69154010-69154010 on Build GRCh38
1735140 c.3879G>T p.W1293C Chr.17: 69153917-69153917 on Build GRCh38
301085 c.3894G>T p.M1298I Chr.17: 69153902-69153902 on Build GRCh38
6589425 c.3836G>A p.S1279N Chr.17: 69153960-69153960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148189434 Chr.17: 69153824-69153824 on Build GRCh38
rs199865559 Chr.17: 69153810-69153810 on Build GRCh38
rs773893541 Chr.17: 69153906-69153906 on Build GRCh38
rs200172999 Chr.17: 69153980-69153980 on Build GRCh38
rs16973656 Chr.17: 69153977-69153977 on Build GRCh38
rs755937158 Chr.17: 69153986-69153986 on Build GRCh38
rs779093748 Chr.17: 69153833-69153833 on Build GRCh38
rs144160248 Chr.17: 69153953-69153953 on Build GRCh38
rs767822848 Chr.17: 69153817-69153817 on Build GRCh38
rs11657280 Chr.17: 69153840-69153840 on Build GRCh38
rs750499120 Chr.17: 69153973-69153973 on Build GRCh38
rs760059626 Chr.17: 69153816-69153816 on Build GRCh38
rs772876949 Chr.17: 69153877-69153877 on Build GRCh38
rs1013210829 Chr.17: 69153828-69153828 on Build GRCh38
rs556543958 Chr.17: 69153937-69153937 on Build GRCh38
rs756471261 Chr.17: 69153823-69153823 on Build GRCh38
rs770470255 Chr.17: 69153905-69153905 on Build GRCh38
rs754919048 Chr.17: 69153850-69153850 on Build GRCh38
rs765281292 Chr.17: 69153971-69153971 on Build GRCh38
rs775793828 Chr.17: 69153917-69153917 on Build GRCh38
rs762450956 Chr.17: 69153798-69153798 on Build GRCh38
rs777515195 Chr.17: 69153990-69153990 on Build GRCh38
rs113881859 Chr.17: 69153875-69153875 on Build GRCh38
rs753094771 Chr.17: 69153822-69153822 on Build GRCh38
rs937091663 Chr.17: 69153876-69153876 on Build GRCh38
rs781192073 Chr.17: 69153978-69153978 on Build GRCh38
rs761135569 Chr.17: 69153933-69153933 on Build GRCh38
rs532320175 Chr.17: 69153870-69153870 on Build GRCh38
rs77718465 Chr.17: 69153995-69153995 on Build GRCh38
rs974631253 Chr.17: 69153802-69153802 on Build GRCh38
rs369143410 Chr.17: 69153910-69153910 on Build GRCh38
rs778354658 Chr.17: 69154003-69154003 on Build GRCh38
rs748904094 Chr.17: 69153883-69153883 on Build GRCh38
rs765830938 Chr.17: 69153803-69153803 on Build GRCh38
rs770558049 Chr.17: 69154001-69154001 on Build GRCh38
rs866283705 Chr.17: 69153938-69153938 on Build GRCh38
rs72852601 Chr.17: 69153831-69153831 on Build GRCh38
rs10491178 Chr.17: 69153832-69153832 on Build GRCh38
rs373370338 Chr.17: 69153912-69153912 on Build GRCh38
rs376127309 Chr.17: 69153861-69153861 on Build GRCh38
rs796080863 Chr.17: 69153871-69153871 on Build GRCh38
rs757521279 Chr.17: 69153954-69153954 on Build GRCh38

View Full Product Details