Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92472004-92472275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
-
UniGene
Hs.367879
Gene Chromosome Location
Chr.15: 92465762-92501117 on Build GRCh38
Transcripts
NM_001301106.1 NM_153040.2 XM_011521257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596380 c.104C>T p.S35F Chr.15: 92472252-92472252 on Build GRCh38
4058008 c.40T>C p.L14L Chr.15: 92472188-92472188 on Build GRCh38
4058007 c.30A>G p.S10S Chr.15: 92472178-92472178 on Build GRCh38
7194864 c.96C>T p.T32T Chr.15: 92472244-92472244 on Build GRCh38
304833 c.47C>T p.P16L Chr.15: 92472195-92472195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571632333 Chr.15: 92472094-92472094 on Build GRCh38
rs1455774 Chr.15: 92472091-92472091 on Build GRCh38
rs988221445 Chr.15: 92472130-92472130 on Build GRCh38
rs188086228 Chr.15: 92472195-92472195 on Build GRCh38
rs745647509 Chr.15: 92472238-92472238 on Build GRCh38
rs779358453 Chr.15: 92472210-92472210 on Build GRCh38
rs571164427 Chr.15: 92472208-92472208 on Build GRCh38
rs763017819 Chr.15: 92472173-92472173 on Build GRCh38
rs995064751 Chr.15: 92472063-92472063 on Build GRCh38
rs772737893 Chr.15: 92472124-92472124 on Build GRCh38
rs1455775 Chr.15: 92472048-92472048 on Build GRCh38
rs770502988 Chr.15: 92472143-92472143 on Build GRCh38
rs763631314 Chr.15: 92472154-92472155 on Build GRCh38
rs925240617 Chr.15: 92472084-92472084 on Build GRCh38
rs117788461 Chr.15: 92472085-92472085 on Build GRCh38
rs769674236 Chr.15: 92472250-92472250 on Build GRCh38
rs115999940 Chr.15: 92472236-92472236 on Build GRCh38
rs147517428 Chr.15: 92472196-92472196 on Build GRCh38
rs771858099 Chr.15: 92472218-92472218 on Build GRCh38
rs770605779 Chr.15: 92472234-92472234 on Build GRCh38
rs146393458 Chr.15: 92472233-92472233 on Build GRCh38
rs530785372 Chr.15: 92472220-92472220 on Build GRCh38
rs969626932 Chr.15: 92472023-92472023 on Build GRCh38
rs777207019 Chr.15: 92472228-92472228 on Build GRCh38
rs887403102 Chr.15: 92472240-92472240 on Build GRCh38
rs746622821 Chr.15: 92472129-92472129 on Build GRCh38
rs1455773 Chr.15: 92472197-92472197 on Build GRCh38
rs776481503 Chr.15: 92472162-92472162 on Build GRCh38
rs748268763 Chr.15: 92472074-92472074 on Build GRCh38
rs769834653 Chr.15: 92472167-92472167 on Build GRCh38
rs747632664 Chr.15: 92472108-92472108 on Build GRCh38
rs142189482 Chr.15: 92472214-92472214 on Build GRCh38
rs759308064 Chr.15: 92472165-92472165 on Build GRCh38
rs750487637 Chr.15: 92472191-92472191 on Build GRCh38
rs767658465 Chr.15: 92472175-92472175 on Build GRCh38
rs771608630 Chr.15: 92472111-92472111 on Build GRCh38
rs983375845 Chr.15: 92472093-92472093 on Build GRCh38
rs200660661 Chr.15: 92472169-92472169 on Build GRCh38

View Full Product Details