Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116768405-116768656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2
Entrez Gene ID
914
Gene Name
CD2 molecule
Gene Aliases
LFA-2, SRBC, T11
UniGene
Hs.523500
Gene Chromosome Location
Chr.1: 116754464-116769229 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7212673 c.810T>C p.A270A Chr.1: 116768537-116768537 on Build GRCh38
7247268 c.788G>A p.R263Q Chr.1: 116768515-116768515 on Build GRCh38
423559 c.742G>T p.E248* Chr.1: 116768469-116768469 on Build GRCh38
4408032 c.799C>A p.Q267K Chr.1: 116768526-116768526 on Build GRCh38
5809409 c.850C>A p.P284T Chr.1: 116768577-116768577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748864256 Chr.1: 116768582-116768582 on Build GRCh38
rs779845333 Chr.1: 116768577-116768577 on Build GRCh38
rs773596778 Chr.1: 116768587-116768587 on Build GRCh38
rs773828485 Chr.1: 116768461-116768461 on Build GRCh38
rs759707410 Chr.1: 116768515-116768515 on Build GRCh38
rs377222693 Chr.1: 116768613-116768613 on Build GRCh38
rs775184596 Chr.1: 116768447-116768447 on Build GRCh38
rs777632148 Chr.1: 116768529-116768529 on Build GRCh38
rs145678592 Chr.1: 116768614-116768614 on Build GRCh38
rs760958788 Chr.1: 116768509-116768509 on Build GRCh38
rs772447791 Chr.1: 116768620-116768620 on Build GRCh38
rs201567293 Chr.1: 116768503-116768503 on Build GRCh38
rs775679910 Chr.1: 116768604-116768604 on Build GRCh38
rs377723619 Chr.1: 116768476-116768476 on Build GRCh38
rs1055585474 Chr.1: 116768460-116768460 on Build GRCh38
rs699738 Chr.1: 116768525-116768525 on Build GRCh38
rs937171505 Chr.1: 116768450-116768450 on Build GRCh38
rs748882774 Chr.1: 116768635-116768635 on Build GRCh38
rs769448924 Chr.1: 116768442-116768442 on Build GRCh38
rs745490266 Chr.1: 116768440-116768440 on Build GRCh38
rs527463622 Chr.1: 116768583-116768583 on Build GRCh38
rs150567561 Chr.1: 116768535-116768535 on Build GRCh38
rs780987566 Chr.1: 116768547-116768547 on Build GRCh38
rs369716425 Chr.1: 116768494-116768494 on Build GRCh38
rs761203896 Chr.1: 116768591-116768591 on Build GRCh38
rs769607172 Chr.1: 116768567-116768567 on Build GRCh38
rs199588776 Chr.1: 116768627-116768627 on Build GRCh38
rs759864783 Chr.1: 116768600-116768600 on Build GRCh38
rs758471774 Chr.1: 116768528-116768528 on Build GRCh38
rs750307503 Chr.1: 116768621-116768621 on Build GRCh38
rs532610592 Chr.1: 116768466-116768466 on Build GRCh38
rs777102347 Chr.1: 116768593-116768593 on Build GRCh38
rs765512102 Chr.1: 116768603-116768603 on Build GRCh38
rs373098399 Chr.1: 116768511-116768511 on Build GRCh38
rs771222747 Chr.1: 116768592-116768592 on Build GRCh38
rs999871668 Chr.1: 116768546-116768546 on Build GRCh38
rs568137617 Chr.1: 116768543-116768543 on Build GRCh38
rs779863841 Chr.1: 116768634-116768634 on Build GRCh38
rs763036877 Chr.1: 116768611-116768611 on Build GRCh38
rs745624462 Chr.1: 116768549-116768549 on Build GRCh38
rs754039794 Chr.1: 116768514-116768514 on Build GRCh38
rs1037996061 Chr.1: 116768436-116768436 on Build GRCh38
rs200585938 Chr.1: 116768497-116768497 on Build GRCh38
rs764171109 Chr.1: 116768612-116768612 on Build GRCh38
rs559951468 Chr.1: 116768553-116768553 on Build GRCh38
rs766698238 Chr.1: 116768512-116768512 on Build GRCh38
rs770420450 Chr.1: 116768451-116768451 on Build GRCh38

View Full Product Details