Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53941955-53942214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG7
Entrez Gene ID
59284
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
UniGene
Hs.631597
Gene Chromosome Location
Chr.19: 53909425-53943944 on Build GRCh38
Transcripts
NM_031896.4 XM_005259124.3 XM_017027093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3693053 c.653G>T p.R218L Chr.19: 53942118-53942118 on Build GRCh38
6558263 c.650A>T p.Y217F Chr.19: 53942115-53942115 on Build GRCh38
79594 c.732C>T p.S244S Chr.19: 53942197-53942197 on Build GRCh38
1723039 c.733delG p.D245fs*10 Chr.19: 53942198-53942198 on Build GRCh38
5567750 c.684G>A p.S228S Chr.19: 53942149-53942149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775015826 Chr.19: 53942098-53942098 on Build GRCh38
rs748447237 Chr.19: 53942167-53942167 on Build GRCh38
rs565456593 Chr.19: 53941999-53941999 on Build GRCh38
rs759102024 Chr.19: 53941998-53941998 on Build GRCh38
rs762464707 Chr.19: 53942190-53942190 on Build GRCh38
rs1035804385 Chr.19: 53942005-53942005 on Build GRCh38
rs750341531 Chr.19: 53942120-53942120 on Build GRCh38
rs772532287 Chr.19: 53942175-53942175 on Build GRCh38
rs768884026 Chr.19: 53942044-53942044 on Build GRCh38
rs111799778 Chr.19: 53941982-53941982 on Build GRCh38
rs145186506 Chr.19: 53942099-53942099 on Build GRCh38
rs551175096 Chr.19: 53942032-53942032 on Build GRCh38
rs749323281 Chr.19: 53942137-53942137 on Build GRCh38
rs762212833 Chr.19: 53942054-53942054 on Build GRCh38
rs769733564 Chr.19: 53942038-53942038 on Build GRCh38
rs772624477 Chr.19: 53942057-53942057 on Build GRCh38
rs773849868 Chr.19: 53942075-53942075 on Build GRCh38
rs557816848 Chr.19: 53942042-53942042 on Build GRCh38
rs547629574 Chr.19: 53942013-53942013 on Build GRCh38
rs147620570 Chr.19: 53942114-53942114 on Build GRCh38
rs771425436 Chr.19: 53942185-53942185 on Build GRCh38
rs756033881 Chr.19: 53942132-53942132 on Build GRCh38
rs951221763 Chr.19: 53941988-53941988 on Build GRCh38
rs750982803 Chr.19: 53941994-53941994 on Build GRCh38
rs773761617 Chr.19: 53942177-53942177 on Build GRCh38
rs768072016 Chr.19: 53941987-53941987 on Build GRCh38
rs774720326 Chr.19: 53942053-53942053 on Build GRCh38
rs755028200 Chr.19: 53942141-53942141 on Build GRCh38
rs781047072 Chr.19: 53942030-53942030 on Build GRCh38
rs947640774 Chr.19: 53942084-53942084 on Build GRCh38
rs115786439 Chr.19: 53942026-53942026 on Build GRCh38
rs149102515 Chr.19: 53942159-53942159 on Build GRCh38
rs751310306 Chr.19: 53942020-53942020 on Build GRCh38
rs536056496 Chr.19: 53941984-53941984 on Build GRCh38
rs576059025 Chr.19: 53942094-53942094 on Build GRCh38
rs376339390 Chr.19: 53942003-53942003 on Build GRCh38
rs866708987 Chr.19: 53942192-53942192 on Build GRCh38
rs555284332 Chr.19: 53942123-53942123 on Build GRCh38
rs368747099 Chr.19: 53942000-53942000 on Build GRCh38
rs781331621 Chr.19: 53942041-53942041 on Build GRCh38
rs537046584 Chr.19: 53942118-53942118 on Build GRCh38
rs777004997 Chr.19: 53942186-53942186 on Build GRCh38
rs911421665 Chr.19: 53942006-53942006 on Build GRCh38
rs541240953 Chr.19: 53942178-53942178 on Build GRCh38
rs761283604 Chr.19: 53942080-53942080 on Build GRCh38
rs762300544 Chr.19: 53941985-53941985 on Build GRCh38
rs372354069 Chr.19: 53942001-53942001 on Build GRCh38
rs748130487 Chr.19: 53942080-53942082 on Build GRCh38

View Full Product Details