Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86306196-86306425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
UniGene
Hs.283322
Gene Chromosome Location
Chr.11: 86302211-86345943 on Build GRCh38
Transcripts
NM_001322404.1 NM_001322407.1 NM_001322409.1 NM_016401.3 NR_024597.1 NR_024598.1 NR_136324.1 XM_017017914.1 XM_017017915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1605316 c.119T>C p.M40T Chr.11: 86306333-86306333 on Build GRCh38
2221645 c.147A>C p.G49G Chr.11: 86306361-86306361 on Build GRCh38
932718 c.95G>A p.S32N Chr.11: 86306309-86306309 on Build GRCh38
7120042 c.58G>A p.E20K Chr.11: 86306272-86306272 on Build GRCh38
4037224 c.88T>C p.Y30H Chr.11: 86306302-86306302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959184928 Chr.11: 86306276-86306276 on Build GRCh38
rs752454830 Chr.11: 86306320-86306320 on Build GRCh38
rs374929851 Chr.11: 86306296-86306296 on Build GRCh38
rs202003795 Chr.11: 86306374-86306374 on Build GRCh38
rs144440797 Chr.11: 86306313-86306313 on Build GRCh38
rs770467937 Chr.11: 86306262-86306262 on Build GRCh38
rs753818305 Chr.11: 86306310-86306310 on Build GRCh38
rs552485887 Chr.11: 86306246-86306246 on Build GRCh38
rs11539213 Chr.11: 86306353-86306353 on Build GRCh38
rs199509279 Chr.11: 86306241-86306241 on Build GRCh38
rs749726674 Chr.11: 86306388-86306388 on Build GRCh38
rs746062689 Chr.11: 86306362-86306362 on Build GRCh38
rs749848152 Chr.11: 86306268-86306268 on Build GRCh38
rs751269890 Chr.11: 86306357-86306357 on Build GRCh38
rs760640958 Chr.11: 86306308-86306308 on Build GRCh38
rs778911484 Chr.11: 86306224-86306224 on Build GRCh38
rs370169348 Chr.11: 86306261-86306261 on Build GRCh38
rs747834214 Chr.11: 86306225-86306225 on Build GRCh38
rs936007437 Chr.11: 86306402-86306402 on Build GRCh38
rs766619246 Chr.11: 86306309-86306309 on Build GRCh38
rs780515938 Chr.11: 86306267-86306267 on Build GRCh38
rs145991257 Chr.11: 86306344-86306344 on Build GRCh38
rs966987196 Chr.11: 86306326-86306326 on Build GRCh38
rs200822371 Chr.11: 86306346-86306346 on Build GRCh38
rs762035750 Chr.11: 86306290-86306290 on Build GRCh38
rs181866288 Chr.11: 86306229-86306229 on Build GRCh38
rs765061366 Chr.11: 86306315-86306315 on Build GRCh38
rs774580967 Chr.11: 86306280-86306280 on Build GRCh38
rs773305668 Chr.11: 86306299-86306299 on Build GRCh38
rs779290255 Chr.11: 86306394-86306394 on Build GRCh38
rs769163297 Chr.11: 86306390-86306390 on Build GRCh38
rs758026918 Chr.11: 86306324-86306324 on Build GRCh38
rs201239774 Chr.11: 86306279-86306279 on Build GRCh38

View Full Product Details