Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90278175-90278444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRDE2
Entrez Gene ID
55051
Gene Name
NRDE-2, necessary for RNA interference, domain containing
Gene Aliases
C14orf102
UniGene
Hs.528131
Gene Chromosome Location
Chr.14: 90274276-90332137 on Build GRCh38
Transcripts
NM_017970.3 XM_011536886.1 XM_017021394.1 XM_017021395.1 XM_017021396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4488930 c.3400C>T p.P1134S Chr.14: 90278431-90278431 on Build GRCh38
5766548 c.3420C>T p.I1140I Chr.14: 90278411-90278411 on Build GRCh38
6729511 c.3387C>T p.A1129A Chr.14: 90278444-90278444 on Build GRCh38
5433048 c.3436G>C p.E1146Q Chr.14: 90278395-90278395 on Build GRCh38
958614 c.3449G>A p.R1150Q Chr.14: 90278382-90278382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039648672 Chr.14: 90278353-90278353 on Build GRCh38
rs773552982 Chr.14: 90278404-90278404 on Build GRCh38
rs773646854 Chr.14: 90278325-90278325 on Build GRCh38
rs772496651 Chr.14: 90278394-90278394 on Build GRCh38
rs567241600 Chr.14: 90278303-90278303 on Build GRCh38
rs569231325 Chr.14: 90278240-90278240 on Build GRCh38
rs983012595 Chr.14: 90278340-90278340 on Build GRCh38
rs781224223 Chr.14: 90278299-90278299 on Build GRCh38
rs369261711 Chr.14: 90278341-90278341 on Build GRCh38
rs537889050 Chr.14: 90278269-90278269 on Build GRCh38
rs917452984 Chr.14: 90278262-90278262 on Build GRCh38
rs748714522 Chr.14: 90278377-90278377 on Build GRCh38
rs778125389 Chr.14: 90278332-90278333 on Build GRCh38
rs761170619 Chr.14: 90278327-90278327 on Build GRCh38
rs779702464 Chr.14: 90278307-90278307 on Build GRCh38
rs369844954 Chr.14: 90278306-90278306 on Build GRCh38
rs751282430 Chr.14: 90278289-90278289 on Build GRCh38
rs145626141 Chr.14: 90278360-90278360 on Build GRCh38
rs762038329 Chr.14: 90278319-90278319 on Build GRCh38
rs775507521 Chr.14: 90278422-90278422 on Build GRCh38
rs756103194 Chr.14: 90278376-90278376 on Build GRCh38
rs45471798 Chr.14: 90278202-90278202 on Build GRCh38
rs747565322 Chr.14: 90278405-90278405 on Build GRCh38
rs754267113 Chr.14: 90278218-90278218 on Build GRCh38
rs776805454 Chr.14: 90278412-90278412 on Build GRCh38
rs781279325 Chr.14: 90278370-90278370 on Build GRCh38
rs10184 Chr.14: 90278328-90278328 on Build GRCh38
rs200016542 Chr.14: 90278288-90278288 on Build GRCh38
rs201104859 Chr.14: 90278420-90278420 on Build GRCh38
rs1051125711 Chr.14: 90278414-90278414 on Build GRCh38
rs755390607 Chr.14: 90278251-90278251 on Build GRCh38
rs765508701 Chr.14: 90278418-90278418 on Build GRCh38
rs768169017 Chr.14: 90278318-90278318 on Build GRCh38
rs146349667 Chr.14: 90278363-90278363 on Build GRCh38
rs1057478946 Chr.14: 90278257-90278257 on Build GRCh38
rs558742926 Chr.14: 90278393-90278393 on Build GRCh38
rs115701639 Chr.14: 90278217-90278217 on Build GRCh38
rs779605631 Chr.14: 90278258-90278258 on Build GRCh38
rs749024128 Chr.14: 90278308-90278308 on Build GRCh38
rs78302825 Chr.14: 90278383-90278383 on Build GRCh38
rs749602262 Chr.14: 90278407-90278408 on Build GRCh38
rs765338598 Chr.14: 90278347-90278347 on Build GRCh38
rs759844752 Chr.14: 90278329-90278329 on Build GRCh38
rs201878322 Chr.14: 90278382-90278382 on Build GRCh38
rs957610251 Chr.14: 90278272-90278272 on Build GRCh38
rs747048338 Chr.14: 90278423-90278423 on Build GRCh38
rs941218122 Chr.14: 90278346-90278346 on Build GRCh38
rs757248022 Chr.14: 90278292-90278292 on Build GRCh38
rs758891089 Chr.14: 90278285-90278285 on Build GRCh38
rs965490533 Chr.14: 90278213-90278213 on Build GRCh38
rs1033211564 Chr.14: 90278271-90278271 on Build GRCh38
rs771398056 Chr.14: 90278410-90278410 on Build GRCh38
rs374089268 Chr.14: 90278320-90278320 on Build GRCh38
rs748554252 Chr.14: 90278386-90278386 on Build GRCh38

View Full Product Details