Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53494474-53494699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
UniGene
Hs.380897
Gene Chromosome Location
Chr.16: 53491028-53504411 on Build GRCh38
Transcripts
NM_001012398.2 NM_001308325.1 NM_022476.3 XM_005256095.4 XM_005256096.4 XM_005256097.4 XM_005256098.4 XM_017023564.1 XM_017023565.1 XM_017023566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7231076 c.416G>T p.R139L Chr.16: 53494604-53494604 on Build GRCh38
4847695 c.442C>T p.H148Y Chr.16: 53494578-53494578 on Build GRCh38
2835017 c.447G>A p.P149P Chr.16: 53494573-53494573 on Build GRCh38
971477 c.415C>T p.R139C Chr.16: 53494605-53494605 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769136279 Chr.16: 53494646-53494646 on Build GRCh38
rs777694925 Chr.16: 53494594-53494594 on Build GRCh38
rs764710208 Chr.16: 53494506-53494506 on Build GRCh38
rs765897827 Chr.16: 53494542-53494542 on Build GRCh38
rs774777157 Chr.16: 53494652-53494652 on Build GRCh38
rs760168557 Chr.16: 53494527-53494527 on Build GRCh38
rs757341232 Chr.16: 53494604-53494604 on Build GRCh38
rs749674014 Chr.16: 53494631-53494631 on Build GRCh38
rs746117124 Chr.16: 53494619-53494619 on Build GRCh38
rs762356281 Chr.16: 53494653-53494653 on Build GRCh38
rs746964570 Chr.16: 53494597-53494597 on Build GRCh38
rs953034676 Chr.16: 53494599-53494599 on Build GRCh38
rs374692428 Chr.16: 53494629-53494629 on Build GRCh38
rs948129533 Chr.16: 53494573-53494573 on Build GRCh38
rs753478965 Chr.16: 53494552-53494552 on Build GRCh38
rs754625474 Chr.16: 53494560-53494560 on Build GRCh38
rs781311887 Chr.16: 53494605-53494605 on Build GRCh38
rs752045477 Chr.16: 53494509-53494509 on Build GRCh38
rs778493299 Chr.16: 53494574-53494574 on Build GRCh38
rs770172988 Chr.16: 53494621-53494621 on Build GRCh38
rs752514939 Chr.16: 53494578-53494578 on Build GRCh38
rs995013998 Chr.16: 53494575-53494575 on Build GRCh38
rs1052650300 Chr.16: 53494651-53494651 on Build GRCh38
rs758135917 Chr.16: 53494589-53494589 on Build GRCh38

View Full Product Details