Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98126432-98126665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2A
Entrez Gene ID
7464
Gene Name
coronin 2A
Gene Aliases
CLIPINB, IR10, WDR2
UniGene
Hs.113094
Gene Chromosome Location
Chr.9: 98120975-98192674 on Build GRCh38
Transcripts
NM_003389.3 NM_052820.3 XM_011518986.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306701 c.1408G>C p.D470H Chr.9: 98126587-98126587 on Build GRCh38
752333 c.1417G>A p.E473K Chr.9: 98126578-98126578 on Build GRCh38
280085 c.1396A>G p.T466A Chr.9: 98126599-98126599 on Build GRCh38
7198027 c.1443T>C p.N481N Chr.9: 98126552-98126552 on Build GRCh38
199466 c.1416C>T p.F472F Chr.9: 98126579-98126579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023050962 Chr.9: 98126538-98126538 on Build GRCh38
rs771489458 Chr.9: 98126584-98126584 on Build GRCh38
rs773410910 Chr.9: 98126502-98126502 on Build GRCh38
rs146742160 Chr.9: 98126535-98126535 on Build GRCh38
rs371084777 Chr.9: 98126561-98126561 on Build GRCh38
rs536898207 Chr.9: 98126562-98126562 on Build GRCh38
rs753607370 Chr.9: 98126546-98126546 on Build GRCh38
rs772352459 Chr.9: 98126615-98126617 on Build GRCh38
rs551331120 Chr.9: 98126454-98126454 on Build GRCh38
rs766971775 Chr.9: 98126530-98126530 on Build GRCh38
rs903117228 Chr.9: 98126472-98126472 on Build GRCh38
rs373005604 Chr.9: 98126585-98126585 on Build GRCh38
rs766872431 Chr.9: 98126534-98126534 on Build GRCh38
rs764950745 Chr.9: 98126624-98126624 on Build GRCh38
rs763453183 Chr.9: 98126507-98126507 on Build GRCh38
rs200947502 Chr.9: 98126569-98126569 on Build GRCh38
rs567921816 Chr.9: 98126555-98126555 on Build GRCh38
rs769878882 Chr.9: 98126571-98126571 on Build GRCh38
rs756903551 Chr.9: 98126621-98126621 on Build GRCh38
rs753553159 Chr.9: 98126617-98126617 on Build GRCh38
rs773618810 Chr.9: 98126578-98126578 on Build GRCh38
rs570721033 Chr.9: 98126616-98126616 on Build GRCh38
rs201721099 Chr.9: 98126493-98126493 on Build GRCh38
rs528218578 Chr.9: 98126531-98126531 on Build GRCh38
rs144845278 Chr.9: 98126642-98126642 on Build GRCh38
rs752115336 Chr.9: 98126614-98126614 on Build GRCh38
rs781087223 Chr.9: 98126568-98126568 on Build GRCh38
rs773485827 Chr.9: 98126581-98126581 on Build GRCh38
rs112540774 Chr.9: 98126609-98126609 on Build GRCh38
rs760280049 Chr.9: 98126615-98126615 on Build GRCh38
rs183116352 Chr.9: 98126598-98126598 on Build GRCh38
rs748663334 Chr.9: 98126531-98126531 on Build GRCh38
rs964036337 Chr.9: 98126481-98126481 on Build GRCh38
rs750243854 Chr.9: 98126639-98126639 on Build GRCh38
rs267602332 Chr.9: 98126579-98126579 on Build GRCh38

View Full Product Details