Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19423767-19424040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER2
Entrez Gene ID
340485
Gene Name
alkaline ceramidase 2
Gene Aliases
ALKCDase2, ASAH3L
UniGene
Hs.41379
Gene Chromosome Location
Chr.9: 19408927-19455173 on Build GRCh38
Transcripts
NM_001010887.2 XM_005251448.3 XM_011517858.1 XM_011517859.2 XM_017014694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1756102 c.19T>C p.Y7H Chr.9: 19423919-19423919 on Build GRCh38
1756101 c.166T>C p.Y56H Chr.9: 19423919-19423919 on Build GRCh38
7256250 c.137C>A p.P46Q Chr.9: 19423890-19423890 on Build GRCh38
1624837 c.13C>T p.R5C Chr.9: 19423913-19423913 on Build GRCh38
6182926 c.109-1G>T p.? Chr.9: 19423861-19423861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145427232 Chr.9: 19423913-19423913 on Build GRCh38
rs750030022 Chr.9: 19423829-19423829 on Build GRCh38
rs762649678 Chr.9: 19423847-19423848 on Build GRCh38
rs900983915 Chr.9: 19423935-19423935 on Build GRCh38
rs746034521 Chr.9: 19423864-19423864 on Build GRCh38
rs768536880 Chr.9: 19423892-19423892 on Build GRCh38
rs369508087 Chr.9: 19423947-19423947 on Build GRCh38
rs556546904 Chr.9: 19423846-19423846 on Build GRCh38
rs772874703 Chr.9: 19423906-19423906 on Build GRCh38
rs986641587 Chr.9: 19423901-19423901 on Build GRCh38
rs749112582 Chr.9: 19423961-19423961 on Build GRCh38
rs374014190 Chr.9: 19423984-19423984 on Build GRCh38
rs368733738 Chr.9: 19423998-19423998 on Build GRCh38
rs749088244 Chr.9: 19423890-19423890 on Build GRCh38
rs778328606 Chr.9: 19423841-19423841 on Build GRCh38
rs373757954 Chr.9: 19423914-19423914 on Build GRCh38
rs771882939 Chr.9: 19423993-19423993 on Build GRCh38
rs761237468 Chr.9: 19423899-19423899 on Build GRCh38
rs771570861 Chr.9: 19423905-19423905 on Build GRCh38
rs757035608 Chr.9: 19423873-19423873 on Build GRCh38
rs80061852 Chr.9: 19423825-19423825 on Build GRCh38
rs768434398 Chr.9: 19423982-19423982 on Build GRCh38
rs544394832 Chr.9: 19423896-19423896 on Build GRCh38
rs370562194 Chr.9: 19423959-19423959 on Build GRCh38
rs200314720 Chr.9: 19423814-19423814 on Build GRCh38
rs139977662 Chr.9: 19423939-19423939 on Build GRCh38
rs761375910 Chr.9: 19423824-19423824 on Build GRCh38
rs74897321 Chr.9: 19423790-19423790 on Build GRCh38
rs776198308 Chr.9: 19424010-19424010 on Build GRCh38
rs750343258 Chr.9: 19423966-19423966 on Build GRCh38
rs775313760 Chr.9: 19423848-19423848 on Build GRCh38
rs371507961 Chr.9: 19423942-19423942 on Build GRCh38
rs896580896 Chr.9: 19424008-19424008 on Build GRCh38
rs765538669 Chr.9: 19423837-19423837 on Build GRCh38
rs781300666 Chr.9: 19423857-19423857 on Build GRCh38
rs780031525 Chr.9: 19423889-19423889 on Build GRCh38
rs759925937 Chr.9: 19423835-19423835 on Build GRCh38
rs762419634 Chr.9: 19424013-19424013 on Build GRCh38
rs371413327 Chr.9: 19423909-19423909 on Build GRCh38
rs377209865 Chr.9: 19424001-19424001 on Build GRCh38
rs367645151 Chr.9: 19423920-19423920 on Build GRCh38
rs111737652 Chr.9: 19423926-19423926 on Build GRCh38
rs191270627 Chr.9: 19423938-19423938 on Build GRCh38
rs201244339 Chr.9: 19423803-19423803 on Build GRCh38
rs746519155 Chr.9: 19424000-19424000 on Build GRCh38
rs753040097 Chr.9: 19423840-19423840 on Build GRCh38
rs764512725 Chr.9: 19423925-19423925 on Build GRCh38
rs146016551 Chr.9: 19423869-19423869 on Build GRCh38
rs756469090 Chr.9: 19423943-19423943 on Build GRCh38
rs974425480 Chr.9: 19423979-19423979 on Build GRCh38
rs267602195 Chr.9: 19423882-19423882 on Build GRCh38
rs78074656 Chr.9: 19424012-19424012 on Build GRCh38
rs757294818 Chr.9: 19423847-19423847 on Build GRCh38
rs752065008 Chr.9: 19423928-19423928 on Build GRCh38
rs773803012 Chr.9: 19423822-19423822 on Build GRCh38
rs556752561 Chr.9: 19423971-19423971 on Build GRCh38
rs748069898 Chr.9: 19423990-19423990 on Build GRCh38

View Full Product Details