Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89583329-89583586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1 XM_017023141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588760 c.504T>A p.D168E Chr.16: 89583450-89583450 on Build GRCh38
4795243 c.558G>A p.R186R Chr.16: 89583504-89583504 on Build GRCh38
3402551 c.569C>T p.T190M Chr.16: 89583515-89583515 on Build GRCh38
6361389 c.571C>T p.Q191* Chr.16: 89583517-89583517 on Build GRCh38
3765961 c.525G>A p.A175A Chr.16: 89583471-89583471 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037178178 Chr.16: 89583371-89583371 on Build GRCh38
rs376596273 Chr.16: 89583393-89583393 on Build GRCh38
rs750997051 Chr.16: 89583522-89583522 on Build GRCh38
rs904047458 Chr.16: 89583488-89583488 on Build GRCh38
rs200338413 Chr.16: 89583518-89583518 on Build GRCh38
rs942720707 Chr.16: 89583469-89583469 on Build GRCh38
rs199770053 Chr.16: 89583557-89583557 on Build GRCh38
rs371142160 Chr.16: 89583556-89583556 on Build GRCh38
rs867565739 Chr.16: 89583519-89583519 on Build GRCh38
rs772025823 Chr.16: 89583558-89583558 on Build GRCh38
rs889868614 Chr.16: 89583454-89583454 on Build GRCh38
rs367690054 Chr.16: 89583567-89583567 on Build GRCh38
rs377149780 Chr.16: 89583356-89583356 on Build GRCh38
rs768369411 Chr.16: 89583388-89583388 on Build GRCh38
rs765782384 Chr.16: 89583520-89583520 on Build GRCh38
rs984824007 Chr.16: 89583346-89583346 on Build GRCh38
rs767096846 Chr.16: 89583543-89583543 on Build GRCh38
rs140031726 Chr.16: 89583470-89583470 on Build GRCh38
rs202128349 Chr.16: 89583515-89583515 on Build GRCh38
rs749103321 Chr.16: 89583480-89583480 on Build GRCh38
rs139566794 Chr.16: 89583484-89583484 on Build GRCh38
rs747903697 Chr.16: 89583453-89583453 on Build GRCh38
rs201466181 Chr.16: 89583516-89583516 on Build GRCh38
rs755741119 Chr.16: 89583550-89583550 on Build GRCh38
rs909742249 Chr.16: 89583354-89583354 on Build GRCh38
rs200477040 Chr.16: 89583497-89583497 on Build GRCh38
rs961668449 Chr.16: 89583535-89583535 on Build GRCh38
rs754442741 Chr.16: 89583539-89583539 on Build GRCh38
rs752266902 Chr.16: 89583544-89583544 on Build GRCh38
rs555386228 Chr.16: 89583494-89583494 on Build GRCh38
rs1022034086 Chr.16: 89583487-89583487 on Build GRCh38
rs748539007 Chr.16: 89583384-89583384 on Build GRCh38
rs933649771 Chr.16: 89583452-89583452 on Build GRCh38
rs761009664 Chr.16: 89583511-89583511 on Build GRCh38
rs369578425 Chr.16: 89583542-89583542 on Build GRCh38
rs780696702 Chr.16: 89583431-89583431 on Build GRCh38
rs150595837 Chr.16: 89583481-89583481 on Build GRCh38
rs368492060 Chr.16: 89583498-89583498 on Build GRCh38
rs749012408 Chr.16: 89583555-89583555 on Build GRCh38
rs1002773627 Chr.16: 89583490-89583490 on Build GRCh38
rs370792220 Chr.16: 89583417-89583417 on Build GRCh38
rs972985694 Chr.16: 89583541-89583541 on Build GRCh38
rs142648138 Chr.16: 89583471-89583471 on Build GRCh38
rs899747565 Chr.16: 89583399-89583399 on Build GRCh38
rs751047820 Chr.16: 89583394-89583394 on Build GRCh38
rs775593028 Chr.16: 89583561-89583561 on Build GRCh38
rs777434423 Chr.16: 89583552-89583552 on Build GRCh38
rs539181727 Chr.16: 89583373-89583373 on Build GRCh38
rs565428038 Chr.16: 89583355-89583355 on Build GRCh38

View Full Product Details