Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44819217-44819480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAM
Entrez Gene ID
4059
Gene Name
basal cell adhesion molecule (Lutheran blood group)
Gene Aliases
AU, CD239, LU, MSK19
UniGene
Hs.625725
Gene Chromosome Location
Chr.19: 44809059-44821421 on Build GRCh38
Transcripts
NM_001013257.2 NM_005581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4467173 c.1475C>T p.P492L Chr.19: 44819347-44819347 on Build GRCh38
3990104 c.1490C>T p.P497L Chr.19: 44819362-44819362 on Build GRCh38
3389124 c.1550G>A p.R517H Chr.19: 44819422-44819422 on Build GRCh38
6350104 c.1545G>A p.L515L Chr.19: 44819417-44819417 on Build GRCh38
5768311 c.1477G>A p.A493T Chr.19: 44819349-44819349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146882795 Chr.19: 44819456-44819456 on Build GRCh38
rs143379896 Chr.19: 44819424-44819424 on Build GRCh38
rs117012738 Chr.19: 44819414-44819414 on Build GRCh38
rs573890694 Chr.19: 44819300-44819300 on Build GRCh38
rs780409275 Chr.19: 44819385-44819385 on Build GRCh38
rs113651096 Chr.19: 44819277-44819277 on Build GRCh38
rs769886489 Chr.19: 44819417-44819417 on Build GRCh38
rs202118823 Chr.19: 44819411-44819411 on Build GRCh38
rs771223969 Chr.19: 44819413-44819413 on Build GRCh38
rs750637876 Chr.19: 44819336-44819336 on Build GRCh38
rs761843311 Chr.19: 44819332-44819332 on Build GRCh38
rs747305308 Chr.19: 44819409-44819409 on Build GRCh38
rs369767093 Chr.19: 44819321-44819321 on Build GRCh38
rs754769675 Chr.19: 44819338-44819338 on Build GRCh38
rs946331764 Chr.19: 44819358-44819358 on Build GRCh38
rs972107158 Chr.19: 44819292-44819292 on Build GRCh38
rs145408767 Chr.19: 44819280-44819280 on Build GRCh38
rs113196154 Chr.19: 44819375-44819375 on Build GRCh38
rs778466916 Chr.19: 44819364-44819364 on Build GRCh38
rs773377986 Chr.19: 44819420-44819420 on Build GRCh38
rs749844885 Chr.19: 44819450-44819450 on Build GRCh38
rs752744214 Chr.19: 44819348-44819348 on Build GRCh38
rs925708575 Chr.19: 44819396-44819396 on Build GRCh38
rs761848492 Chr.19: 44819415-44819415 on Build GRCh38
rs763227493 Chr.19: 44819421-44819421 on Build GRCh38
rs778471984 Chr.19: 44819446-44819446 on Build GRCh38
rs934382784 Chr.19: 44819412-44819412 on Build GRCh38
rs148391498 Chr.19: 44819367-44819367 on Build GRCh38
rs772377922 Chr.19: 44819368-44819368 on Build GRCh38
rs764525698 Chr.19: 44819325-44819325 on Build GRCh38
rs200226990 Chr.19: 44819346-44819346 on Build GRCh38
rs539905238 Chr.19: 44819422-44819422 on Build GRCh38
rs756765097 Chr.19: 44819363-44819363 on Build GRCh38
rs776695346 Chr.19: 44819326-44819326 on Build GRCh38
rs367879811 Chr.19: 44819361-44819361 on Build GRCh38
rs372072867 Chr.19: 44819423-44819423 on Build GRCh38
rs773032810 Chr.19: 44819302-44819307 on Build GRCh38
rs1002740847 Chr.19: 44819261-44819261 on Build GRCh38
rs927328751 Chr.19: 44819279-44819279 on Build GRCh38
rs371714918 Chr.19: 44819333-44819333 on Build GRCh38
rs373734560 Chr.19: 44819349-44819349 on Build GRCh38
rs761156550 Chr.19: 44819322-44819322 on Build GRCh38
rs973516903 Chr.19: 44819273-44819273 on Build GRCh38
rs528037775 Chr.19: 44819460-44819460 on Build GRCh38
rs747288346 Chr.19: 44819380-44819381 on Build GRCh38
rs367650807 Chr.19: 44819235-44819235 on Build GRCh38

View Full Product Details