Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159774993-159775264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAT2
Entrez Gene ID
39
Gene Name
acetyl-CoA acetyltransferase 2
Gene Aliases
-
UniGene
Hs.571037
Gene Chromosome Location
Chr.6: 159761957-159779112 on Build GRCh38
Transcripts
NM_001303253.1 NM_005891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3023321 c.493G>T p.E165* Chr.6: 159775172-159775172 on Build GRCh38
7203299 c.623A>T p.D208V Chr.6: 159775215-159775215 on Build GRCh38
158842 c.550C>G p.L184V Chr.6: 159775229-159775229 on Build GRCh38
4910666 c.584C>T p.A195V Chr.6: 159775263-159775263 on Build GRCh38
7203298 c.536A>T p.D179V Chr.6: 159775215-159775215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759956506 Chr.6: 159775226-159775226 on Build GRCh38
rs774923207 Chr.6: 159775184-159775184 on Build GRCh38
rs377702862 Chr.6: 159775177-159775177 on Build GRCh38
rs772195941 Chr.6: 159775191-159775191 on Build GRCh38
rs910357883 Chr.6: 159775098-159775098 on Build GRCh38
rs773999963 Chr.6: 159775238-159775238 on Build GRCh38
rs371307389 Chr.6: 159775205-159775205 on Build GRCh38
rs181382189 Chr.6: 159775129-159775129 on Build GRCh38
rs756861222 Chr.6: 159775126-159775126 on Build GRCh38
rs1000550825 Chr.6: 159775040-159775040 on Build GRCh38
rs755952672 Chr.6: 159775162-159775162 on Build GRCh38
rs943243618 Chr.6: 159775115-159775115 on Build GRCh38
rs990120457 Chr.6: 159775095-159775095 on Build GRCh38
rs1032067892 Chr.6: 159775057-159775057 on Build GRCh38
rs779334826 Chr.6: 159775179-159775179 on Build GRCh38
rs753057472 Chr.6: 159775130-159775135 on Build GRCh38
rs749131115 Chr.6: 159775170-159775170 on Build GRCh38
rs200038002 Chr.6: 159775142-159775142 on Build GRCh38
rs978089329 Chr.6: 159775031-159775031 on Build GRCh38
rs1028144499 Chr.6: 159775240-159775240 on Build GRCh38
rs779925783 Chr.6: 159775165-159775165 on Build GRCh38
rs1019737660 Chr.6: 159775150-159775150 on Build GRCh38
rs771382288 Chr.6: 159775220-159775220 on Build GRCh38
rs761277497 Chr.6: 159775210-159775210 on Build GRCh38
rs892223614 Chr.6: 159775070-159775070 on Build GRCh38
rs571822479 Chr.6: 159775036-159775037 on Build GRCh38
rs1047395793 Chr.6: 159775143-159775143 on Build GRCh38
rs769728018 Chr.6: 159775139-159775140 on Build GRCh38
rs534790505 Chr.6: 159775183-159775183 on Build GRCh38
rs777100728 Chr.6: 159775224-159775224 on Build GRCh38

View Full Product Details