Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46695121-46695382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERK
Entrez Gene ID
64781
Gene Name
ceramide kinase
Gene Aliases
LK4, dA59H18.2, dA59H18.3, hCERK
UniGene
Hs.200668
Gene Chromosome Location
Chr.22: 46684410-46738255 on Build GRCh38
Transcripts
NM_022766.5 XM_017028909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3748770 c.1044G>T p.R348R Chr.22: 46695215-46695215 on Build GRCh38
3964406 c.1043G>T p.R348L Chr.22: 46695216-46695216 on Build GRCh38
7415903 c.1033A>G p.K345E Chr.22: 46695226-46695226 on Build GRCh38
4996083 c.1027G>A p.D343N Chr.22: 46695232-46695232 on Build GRCh38
6627305 c.1024A>G p.R342G Chr.22: 46695235-46695235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754115109 Chr.22: 46695253-46695253 on Build GRCh38
rs768758197 Chr.22: 46695224-46695224 on Build GRCh38
rs781641885 Chr.22: 46695179-46695179 on Build GRCh38
rs9616098 Chr.22: 46695334-46695334 on Build GRCh38
rs36211081 Chr.22: 46695246-46695246 on Build GRCh38
rs770637394 Chr.22: 46695329-46695329 on Build GRCh38
rs781190421 Chr.22: 46695288-46695288 on Build GRCh38
rs115160162 Chr.22: 46695361-46695361 on Build GRCh38
rs769366380 Chr.22: 46695308-46695308 on Build GRCh38
rs747007748 Chr.22: 46695169-46695169 on Build GRCh38
rs778071127 Chr.22: 46695205-46695205 on Build GRCh38
rs115008707 Chr.22: 46695195-46695195 on Build GRCh38
rs772946630 Chr.22: 46695232-46695232 on Build GRCh38
rs758067862 Chr.22: 46695154-46695154 on Build GRCh38
rs776692410 Chr.22: 46695226-46695226 on Build GRCh38
rs368437490 Chr.22: 46695249-46695249 on Build GRCh38
rs200063352 Chr.22: 46695189-46695189 on Build GRCh38
rs371610703 Chr.22: 46695311-46695311 on Build GRCh38
rs762461739 Chr.22: 46695237-46695237 on Build GRCh38
rs577496966 Chr.22: 46695217-46695217 on Build GRCh38
rs749406399 Chr.22: 46695213-46695213 on Build GRCh38
rs751960449 Chr.22: 46695245-46695245 on Build GRCh38
rs769778006 Chr.22: 46695179-46695180 on Build GRCh38
rs372234931 Chr.22: 46695172-46695172 on Build GRCh38
rs1054290836 Chr.22: 46695356-46695356 on Build GRCh38
rs148368178 Chr.22: 46695216-46695216 on Build GRCh38
rs769900283 Chr.22: 46695285-46695285 on Build GRCh38
rs561978624 Chr.22: 46695353-46695353 on Build GRCh38
rs749140427 Chr.22: 46695244-46695246 on Build GRCh38
rs779965424 Chr.22: 46695188-46695189 on Build GRCh38
rs200157244 Chr.22: 46695230-46695230 on Build GRCh38
rs747993114 Chr.22: 46695297-46695297 on Build GRCh38
rs765992916 Chr.22: 46695244-46695244 on Build GRCh38
rs746854966 Chr.22: 46695277-46695277 on Build GRCh38
rs749983775 Chr.22: 46695159-46695159 on Build GRCh38
rs777349994 Chr.22: 46695171-46695171 on Build GRCh38
rs766687294 Chr.22: 46695166-46695166 on Build GRCh38
rs749102053 Chr.22: 46695359-46695359 on Build GRCh38
rs762493196 Chr.22: 46695317-46695317 on Build GRCh38
rs756761355 Chr.22: 46695229-46695229 on Build GRCh38
rs759907770 Chr.22: 46695352-46695352 on Build GRCh38
rs556758428 Chr.22: 46695248-46695248 on Build GRCh38
rs368958311 Chr.22: 46695357-46695357 on Build GRCh38
rs757605254 Chr.22: 46695256-46695256 on Build GRCh38

View Full Product Details