Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 541410-541635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC34
Entrez Gene ID
997
Gene Name
cell division cycle 34
Gene Aliases
E2-CDC34, UBC3, UBCH3, UBE2R1
UniGene
Hs.514997
Gene Chromosome Location
Chr.19: 531720-542088 on Build GRCh38
Transcripts
NM_004359.1 XM_006722952.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960432 c.660C>T p.A220A Chr.19: 541501-541501 on Build GRCh38
6624221 c.702G>A p.E234E Chr.19: 541543-541543 on Build GRCh38
4953714 c.708C>A p.S236S Chr.19: 541549-541549 on Build GRCh38
4670672 c.634G>A p.E212K Chr.19: 541475-541475 on Build GRCh38
4581379 c.622G>C p.D208H Chr.19: 541463-541463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034014803 Chr.19: 541482-541482 on Build GRCh38
rs556391590 Chr.19: 541439-541439 on Build GRCh38
rs748408800 Chr.19: 541459-541461 on Build GRCh38
rs16990650 Chr.19: 541520-541520 on Build GRCh38
rs777308193 Chr.19: 541513-541513 on Build GRCh38
rs745626145 Chr.19: 541484-541484 on Build GRCh38
rs986506089 Chr.19: 541592-541592 on Build GRCh38
rs148173026 Chr.19: 541429-541429 on Build GRCh38
rs767929636 Chr.19: 541432-541432 on Build GRCh38
rs754528124 Chr.19: 541558-541558 on Build GRCh38
rs542019154 Chr.19: 541459-541459 on Build GRCh38
rs772271528 Chr.19: 541467-541469 on Build GRCh38
rs370735978 Chr.19: 541574-541574 on Build GRCh38
rs376816532 Chr.19: 541572-541572 on Build GRCh38
rs368818872 Chr.19: 541590-541590 on Build GRCh38
rs757087028 Chr.19: 541475-541475 on Build GRCh38
rs907074961 Chr.19: 541607-541607 on Build GRCh38
rs369872688 Chr.19: 541598-541598 on Build GRCh38
rs751666566 Chr.19: 541470-541470 on Build GRCh38
rs925159826 Chr.19: 541611-541611 on Build GRCh38
rs146221392 Chr.19: 541540-541540 on Build GRCh38
rs150670250 Chr.19: 541456-541456 on Build GRCh38
rs767629155 Chr.19: 541537-541537 on Build GRCh38
rs746366208 Chr.19: 541489-541491 on Build GRCh38
rs773664509 Chr.19: 541500-541500 on Build GRCh38
rs750335335 Chr.19: 541539-541539 on Build GRCh38
rs768025971 Chr.19: 541453-541453 on Build GRCh38
rs761024725 Chr.19: 541505-541506 on Build GRCh38
rs771514074 Chr.19: 541502-541502 on Build GRCh38
rs761398120 Chr.19: 541501-541501 on Build GRCh38
rs568742634 Chr.19: 541514-541514 on Build GRCh38
rs140384237 Chr.19: 541536-541536 on Build GRCh38
rs537733016 Chr.19: 541586-541586 on Build GRCh38
rs762703606 Chr.19: 541443-541443 on Build GRCh38
rs892526789 Chr.19: 541593-541593 on Build GRCh38
rs550012804 Chr.19: 541496-541496 on Build GRCh38
rs781133107 Chr.19: 541483-541483 on Build GRCh38
rs748875983 Chr.19: 541494-541494 on Build GRCh38
rs765684353 Chr.19: 541519-541519 on Build GRCh38
rs775660862 Chr.19: 541600-541600 on Build GRCh38
rs575169224 Chr.19: 541614-541617 on Build GRCh38
rs531877075 Chr.19: 541493-541493 on Build GRCh38
rs762501839 Chr.19: 541596-541598 on Build GRCh38
rs772915346 Chr.19: 541577-541577 on Build GRCh38
rs377177218 Chr.19: 541441-541441 on Build GRCh38
rs201584025 Chr.19: 541442-541442 on Build GRCh38
rs200761288 Chr.19: 541463-541463 on Build GRCh38
rs1001414844 Chr.19: 541613-541613 on Build GRCh38
rs369427490 Chr.19: 541435-541435 on Build GRCh38
rs563776588 Chr.19: 541487-541487 on Build GRCh38
rs201715065 Chr.19: 541523-541523 on Build GRCh38
rs141634653 Chr.19: 541465-541465 on Build GRCh38
rs771538108 Chr.19: 541513-541514 on Build GRCh38
rs768762379 Chr.19: 541525-541525 on Build GRCh38
rs369990220 Chr.19: 541460-541460 on Build GRCh38
rs376164577 Chr.19: 541546-541546 on Build GRCh38
rs141921412 Chr.19: 541438-541438 on Build GRCh38
rs751546350 Chr.19: 541533-541533 on Build GRCh38
rs375818778 Chr.19: 541434-541434 on Build GRCh38
rs778649386 Chr.19: 541568-541568 on Build GRCh38
rs746373006 Chr.19: 541594-541594 on Build GRCh38
rs1034276097 Chr.19: 541614-541614 on Build GRCh38
rs777711920 Chr.19: 541466-541466 on Build GRCh38
rs779289790 Chr.19: 541431-541431 on Build GRCh38
rs191947256 Chr.19: 541599-541599 on Build GRCh38
rs747570195 Chr.19: 541573-541573 on Build GRCh38
rs147105441 Chr.19: 541428-541428 on Build GRCh38
rs903828650 Chr.19: 541564-541564 on Build GRCh38
rs774852758 Chr.19: 541531-541532 on Build GRCh38
rs1039934031 Chr.19: 541555-541555 on Build GRCh38
rs139926290 Chr.19: 541462-541462 on Build GRCh38

View Full Product Details